Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8261483-8261756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS4
Entrez Gene ID
79603
Gene Name
ceramide synthase 4
Gene Aliases
LASS4, Trh1
UniGene
Hs.515111
Gene Chromosome Location
Chr.19: 8209329-8262433 on Build GRCh38
Transcripts
NM_024552.2 XM_011528290.2 XM_011528291.2 XM_011528292.2 XM_011528293.2 XM_011528294.2 XM_011528295.2 XM_017027303.1 XM_017027304.1 XM_017027305.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6153357 c.912C>G p.F304L Chr.19: 8261751-8261751 on Build GRCh38
6443870 c.902G>A p.G301D Chr.19: 8261741-8261741 on Build GRCh38
6195599 c.871G>C p.E291Q Chr.19: 8261710-8261710 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs935909866 Chr.19: 8261601-8261601 on Build GRCh38
rs371201164 Chr.19: 8261638-8261638 on Build GRCh38
rs192059550 Chr.19: 8261642-8261642 on Build GRCh38
rs757498502 Chr.19: 8261657-8261657 on Build GRCh38
rs993004648 Chr.19: 8261523-8261523 on Build GRCh38
rs142319862 Chr.19: 8261710-8261710 on Build GRCh38
rs569506788 Chr.19: 8261545-8261545 on Build GRCh38
rs769384251 Chr.19: 8261690-8261690 on Build GRCh38
rs777892996 Chr.19: 8261663-8261665 on Build GRCh38
rs538159802 Chr.19: 8261562-8261562 on Build GRCh38
rs555902738 Chr.19: 8261528-8261528 on Build GRCh38
rs187336194 Chr.19: 8261625-8261625 on Build GRCh38
rs114452930 Chr.19: 8261724-8261724 on Build GRCh38
rs1005149817 Chr.19: 8261711-8261711 on Build GRCh38
rs750126620 Chr.19: 8261645-8261645 on Build GRCh38
rs200811941 Chr.19: 8261665-8261665 on Build GRCh38
rs199736581 Chr.19: 8261652-8261652 on Build GRCh38
rs996725416 Chr.19: 8261563-8261564 on Build GRCh38
rs751521709 Chr.19: 8261712-8261712 on Build GRCh38
rs111265727 Chr.19: 8261555-8261555 on Build GRCh38
rs772451622 Chr.19: 8261662-8261662 on Build GRCh38
rs1015494223 Chr.19: 8261506-8261506 on Build GRCh38
rs1008574956 Chr.19: 8261730-8261730 on Build GRCh38
rs1043496653 Chr.19: 8261703-8261703 on Build GRCh38
rs998274814 Chr.19: 8261612-8261612 on Build GRCh38
rs1021938013 Chr.19: 8261537-8261537 on Build GRCh38
rs555105242 Chr.19: 8261569-8261569 on Build GRCh38
rs745378071 Chr.19: 8261673-8261674 on Build GRCh38
rs773087994 Chr.19: 8261702-8261702 on Build GRCh38
rs370319528 Chr.19: 8261718-8261718 on Build GRCh38
rs1049072573 Chr.19: 8261684-8261684 on Build GRCh38
rs746180647 Chr.19: 8261661-8261661 on Build GRCh38
rs756307675 Chr.19: 8261653-8261653 on Build GRCh38
rs960874982 Chr.19: 8261515-8261515 on Build GRCh38
rs576236985 Chr.19: 8261677-8261677 on Build GRCh38
rs955373241 Chr.19: 8261571-8261571 on Build GRCh38
rs944517844 Chr.19: 8261643-8261643 on Build GRCh38
rs1024016307 Chr.19: 8261529-8261529 on Build GRCh38
rs755943622 Chr.19: 8261620-8261620 on Build GRCh38
rs778197020 Chr.19: 8261649-8261650 on Build GRCh38
rs548486184 Chr.19: 8261594-8261594 on Build GRCh38
rs767508547 Chr.19: 8261560-8261560 on Build GRCh38
rs201536694 Chr.19: 8261660-8261660 on Build GRCh38
rs770943490 Chr.19: 8261729-8261730 on Build GRCh38
rs61729807 Chr.19: 8261727-8261727 on Build GRCh38
rs780475833 Chr.19: 8261664-8261664 on Build GRCh38
rs945072249 Chr.19: 8261683-8261683 on Build GRCh38
rs776050248 Chr.19: 8261666-8261666 on Build GRCh38
rs986677315 Chr.19: 8261587-8261587 on Build GRCh38
rs764284889 Chr.19: 8261719-8261719 on Build GRCh38
rs769311109 Chr.19: 8261681-8261686 on Build GRCh38
rs913075964 Chr.19: 8261636-8261636 on Build GRCh38
rs202234195 Chr.19: 8261646-8261646 on Build GRCh38
rs777999244 Chr.19: 8261654-8261654 on Build GRCh38

View Full Product Details