Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 16154640-16154897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC6
Entrez Gene ID
368
Gene Name
ATP binding cassette subfamily C member 6
Gene Aliases
ABC34, ARA, EST349056, GACI2, MLP1, MOAT-E, MOATE, MRP6, PXE, PXE1, URG7
UniGene
Hs.442182
Gene Chromosome Location
Chr.16: 16149565-16223616 on Build GRCh38
Transcripts
NM_001171.5 XM_011522479.2 XM_011522480.1 XM_011522481.2 XM_017023212.1 XM_017023213.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1376140 c.4080C>T p.L1360L Chr.16: 16154756-16154756 on Build GRCh38
6849918 c.4037C>T p.P1346L Chr.16: 16154877-16154877 on Build GRCh38
6370909 c.4157G>T p.S1386I Chr.16: 16154679-16154679 on Build GRCh38
3356803 c.4133T>C p.V1378A Chr.16: 16154703-16154703 on Build GRCh38
6393098 c.4126G>T p.E1376* Chr.16: 16154710-16154710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766760351 Chr.16: 16154840-16154840 on Build GRCh38
rs763399476 Chr.16: 16154839-16154839 on Build GRCh38
rs199668617 Chr.16: 16154732-16154732 on Build GRCh38
rs764446055 Chr.16: 16154674-16154674 on Build GRCh38
rs1012629344 Chr.16: 16154810-16154810 on Build GRCh38
rs956434099 Chr.16: 16154864-16154864 on Build GRCh38
rs748103958 Chr.16: 16154710-16154710 on Build GRCh38
rs368258006 Chr.16: 16154696-16154696 on Build GRCh38
rs759717857 Chr.16: 16154845-16154845 on Build GRCh38
rs771581942 Chr.16: 16154741-16154741 on Build GRCh38
rs750674262 Chr.16: 16154777-16154777 on Build GRCh38
rs758486813 Chr.16: 16154780-16154780 on Build GRCh38
rs200252305 Chr.16: 16154857-16154857 on Build GRCh38
rs61174588 Chr.16: 16154812-16154812 on Build GRCh38
rs578175155 Chr.16: 16154799-16154799 on Build GRCh38
rs779667151 Chr.16: 16154816-16154816 on Build GRCh38
rs200834294 Chr.16: 16154735-16154735 on Build GRCh38
rs545644418 Chr.16: 16154801-16154801 on Build GRCh38
rs141821068 Chr.16: 16154786-16154786 on Build GRCh38
rs63750428 Chr.16: 16154767-16154767 on Build GRCh38
rs765472331 Chr.16: 16154772-16154772 on Build GRCh38
rs766707250 Chr.16: 16154783-16154783 on Build GRCh38
rs767444769 Chr.16: 16154847-16154847 on Build GRCh38
rs60810627 Chr.16: 16154823-16154823 on Build GRCh38
rs201275608 Chr.16: 16154766-16154766 on Build GRCh38
rs57734166 Chr.16: 16154815-16154815 on Build GRCh38
rs72664288 Chr.16: 16154788-16154788 on Build GRCh38
rs770201316 Chr.16: 16154830-16154830 on Build GRCh38
rs560660056 Chr.16: 16154806-16154806 on Build GRCh38
rs757074599 Chr.16: 16154863-16154863 on Build GRCh38
rs63751111 Chr.16: 16154873-16154873 on Build GRCh38
rs759123370 Chr.16: 16154662-16154662 on Build GRCh38
rs944132085 Chr.16: 16154682-16154682 on Build GRCh38
rs764635677 Chr.16: 16154763-16154763 on Build GRCh38
rs766971195 Chr.16: 16154665-16154665 on Build GRCh38
rs754267653 Chr.16: 16154685-16154685 on Build GRCh38
rs750243019 Chr.16: 16154877-16154877 on Build GRCh38
rs777894623 Chr.16: 16154800-16154800 on Build GRCh38
rs749136865 Chr.16: 16154807-16154807 on Build GRCh38
rs373736094 Chr.16: 16154672-16154672 on Build GRCh38
rs757540398 Chr.16: 16154687-16154687 on Build GRCh38
rs750573670 Chr.16: 16154705-16154705 on Build GRCh38
rs775998775 Chr.16: 16154671-16154671 on Build GRCh38
rs747334212 Chr.16: 16154664-16154665 on Build GRCh38
rs143181260 Chr.16: 16154829-16154829 on Build GRCh38
rs60285147 Chr.16: 16154731-16154731 on Build GRCh38
rs777835756 Chr.16: 16154728-16154728 on Build GRCh38
rs58695352 Chr.16: 16154755-16154755 on Build GRCh38
rs371122482 Chr.16: 16154706-16154706 on Build GRCh38
rs751796653 Chr.16: 16154843-16154843 on Build GRCh38
rs375386682 Chr.16: 16154729-16154729 on Build GRCh38
rs369909327 Chr.16: 16154834-16154834 on Build GRCh38
rs534017491 Chr.16: 16154660-16154660 on Build GRCh38
rs371489110 Chr.16: 16154809-16154809 on Build GRCh38
rs72664237 Chr.16: 16154732-16154732 on Build GRCh38
rs63750018 Chr.16: 16154776-16154776 on Build GRCh38
rs2066738 Chr.16: 16154824-16154824 on Build GRCh38
rs139978668 Chr.16: 16154750-16154750 on Build GRCh38
rs777218739 Chr.16: 16154826-16154826 on Build GRCh38
rs756191351 Chr.16: 16154713-16154713 on Build GRCh38

View Full Product Details