Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29066675-29066926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT8
Entrez Gene ID
10694
Gene Name
chaperonin containing TCP1 subunit 8
Gene Aliases
C21orf112, Cctq, D21S246, PRED71
UniGene
Hs.125113
Gene Chromosome Location
Chr.21: 29056322-29073797 on Build GRCh38
Transcripts
NM_001282907.1 NM_001282908.1 NM_001282909.1 NM_006585.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
243547 c.599A>G p.D200G Chr.21: 29066741-29066741 on Build GRCh38
6287045 c.544C>A p.L182I Chr.21: 29066909-29066909 on Build GRCh38
4101024 c.562+1G>A p.? Chr.21: 29066890-29066890 on Build GRCh38
7360169 c.583G>A p.G195S Chr.21: 29066757-29066757 on Build GRCh38
4944600 c.597T>C p.V199V Chr.21: 29066743-29066743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760352658 Chr.21: 29066712-29066712 on Build GRCh38
rs775548415 Chr.21: 29066812-29066812 on Build GRCh38
rs373606062 Chr.21: 29066865-29066865 on Build GRCh38
rs771509816 Chr.21: 29066796-29066796 on Build GRCh38
rs759533265 Chr.21: 29066802-29066802 on Build GRCh38
rs905420592 Chr.21: 29066736-29066736 on Build GRCh38
rs1032322384 Chr.21: 29066815-29066815 on Build GRCh38
rs781006894 Chr.21: 29066882-29066882 on Build GRCh38
rs988188508 Chr.21: 29066841-29066841 on Build GRCh38
rs761608154 Chr.21: 29066840-29066840 on Build GRCh38
rs375877034 Chr.21: 29066776-29066776 on Build GRCh38
rs775059643 Chr.21: 29066799-29066799 on Build GRCh38
rs778245205 Chr.21: 29066784-29066784 on Build GRCh38
rs1023763395 Chr.21: 29066709-29066709 on Build GRCh38
rs749877026 Chr.21: 29066759-29066759 on Build GRCh38
rs866862449 Chr.21: 29066756-29066756 on Build GRCh38
rs759948624 Chr.21: 29066878-29066878 on Build GRCh38
rs878885914 Chr.21: 29066769-29066769 on Build GRCh38
rs773886434 Chr.21: 29066798-29066798 on Build GRCh38
rs533997184 Chr.21: 29066848-29066848 on Build GRCh38
rs765144024 Chr.21: 29066863-29066863 on Build GRCh38
rs764327507 Chr.21: 29066821-29066821 on Build GRCh38
rs749676122 Chr.21: 29066794-29066794 on Build GRCh38
rs137862783 Chr.21: 29066891-29066891 on Build GRCh38
rs200522803 Chr.21: 29066892-29066892 on Build GRCh38
rs763994165 Chr.21: 29066728-29066728 on Build GRCh38
rs375033335 Chr.21: 29066785-29066788 on Build GRCh38
rs370516107 Chr.21: 29066819-29066819 on Build GRCh38
rs140880362 Chr.21: 29066783-29066786 on Build GRCh38
rs778824426 Chr.21: 29066758-29066758 on Build GRCh38
rs748001135 Chr.21: 29066824-29066827 on Build GRCh38
rs753653342 Chr.21: 29066747-29066747 on Build GRCh38
rs771674921 Chr.21: 29066872-29066872 on Build GRCh38
rs768216110 Chr.21: 29066780-29066780 on Build GRCh38
rs751063625 Chr.21: 29066875-29066875 on Build GRCh38
rs11088108 Chr.21: 29066708-29066708 on Build GRCh38
rs1001540054 Chr.21: 29066808-29066808 on Build GRCh38
rs759002697 Chr.21: 29066836-29066836 on Build GRCh38
rs765864738 Chr.21: 29066870-29066870 on Build GRCh38
rs757144494 Chr.21: 29066754-29066754 on Build GRCh38
rs754281981 Chr.21: 29066807-29066807 on Build GRCh38
rs201031013 Chr.21: 29066772-29066772 on Build GRCh38
rs779536062 Chr.21: 29066902-29066902 on Build GRCh38
rs757815887 Chr.21: 29066866-29066866 on Build GRCh38
rs778937654 Chr.21: 29066806-29066808 on Build GRCh38
rs558654266 Chr.21: 29066775-29066775 on Build GRCh38
rs753560529 Chr.21: 29066824-29066824 on Build GRCh38
rs765513230 Chr.21: 29066879-29066879 on Build GRCh38

View Full Product Details