Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20777162-20777407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM3
Entrez Gene ID
6296
Gene Name
acyl-CoA synthetase medium-chain family member 3
Gene Aliases
SA, SAH
UniGene
Hs.706754
Gene Chromosome Location
Chr.16: 20763990-20797157 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
968135 c.460C>G p.L154V Chr.16: 20777402-20777402 on Build GRCh38
5732688 c.457C>A p.Q153K Chr.16: 20777399-20777399 on Build GRCh38
6464254 c.436G>T p.V146F Chr.16: 20777378-20777378 on Build GRCh38
1580410 c.449G>A p.G150E Chr.16: 20777391-20777391 on Build GRCh38
1588318 c.460C>G p.L154V Chr.16: 20777402-20777402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973291793 Chr.16: 20777249-20777249 on Build GRCh38
rs772217945 Chr.16: 20777381-20777381 on Build GRCh38
rs781120681 Chr.16: 20777340-20777340 on Build GRCh38
rs749256255 Chr.16: 20777350-20777350 on Build GRCh38
rs754860763 Chr.16: 20777330-20777330 on Build GRCh38
rs778986438 Chr.16: 20777366-20777366 on Build GRCh38
rs761137788 Chr.16: 20777387-20777387 on Build GRCh38
rs1021676222 Chr.16: 20777273-20777273 on Build GRCh38
rs771106487 Chr.16: 20777352-20777352 on Build GRCh38
rs746148438 Chr.16: 20777380-20777380 on Build GRCh38
rs1000676491 Chr.16: 20777335-20777335 on Build GRCh38
rs13306611 Chr.16: 20777240-20777240 on Build GRCh38
rs113476421 Chr.16: 20777267-20777267 on Build GRCh38
rs751532131 Chr.16: 20777345-20777345 on Build GRCh38
rs967829188 Chr.16: 20777275-20777275 on Build GRCh38
rs367601507 Chr.16: 20777244-20777244 on Build GRCh38
rs752449738 Chr.16: 20777372-20777373 on Build GRCh38
rs185502355 Chr.16: 20777187-20777187 on Build GRCh38
rs775951468 Chr.16: 20777383-20777383 on Build GRCh38

View Full Product Details