Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 96879750-96880011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERAP1
Entrez Gene ID
51752
Gene Name
endoplasmic reticulum aminopeptidase 1
Gene Aliases
A-LAP, ALAP, APPILS, ARTS-1, ARTS1, ERAAP, ERAAP1, PILS-AP, PILSAP
UniGene
Hs.666524
Gene Chromosome Location
Chr.5: 96759245-96935983 on Build GRCh38
Transcripts
XM_011543480.2 XM_011543481.2 XM_011543484.2 XM_011543485.2 XM_011543486.2 XM_017009580.1 XM_017009581.1 XM_011543544.1

Gene Symbol
ERAP2
Entrez Gene ID
64167
Gene Name
endoplasmic reticulum aminopeptidase 2
Gene Aliases
L-RAP, LRAP
UniGene
Hs.482910
Gene Chromosome Location
Chr.5: 96875940-96919703 on Build GRCh38
Transcripts
XM_011543480.2 XM_011543481.2 XM_011543484.2 XM_011543485.2 XM_011543486.2 XM_017009580.1 XM_017009581.1 XM_011543544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4648936 c.292G>A p.E98K Chr.5: 96879977-96879977 on Build GRCh38
3429785 c.114C>T p.F38F Chr.5: 96879799-96879799 on Build GRCh38
4137902 c.226C>T p.L76F Chr.5: 96879911-96879911 on Build GRCh38
1071472 c.288G>T p.K96N Chr.5: 96879973-96879973 on Build GRCh38
6565776 c.99C>T p.C33C Chr.5: 96879784-96879784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764675283 Chr.5: 96879860-96879860 on Build GRCh38
rs373559640 Chr.5: 96879980-96879980 on Build GRCh38
rs751559114 Chr.5: 96879925-96879925 on Build GRCh38
rs144285538 Chr.5: 96879799-96879799 on Build GRCh38
rs769082342 Chr.5: 96879774-96879774 on Build GRCh38
rs573034708 Chr.5: 96879773-96879773 on Build GRCh38
rs905153527 Chr.5: 96879954-96879954 on Build GRCh38
rs562967580 Chr.5: 96879915-96879915 on Build GRCh38
rs752426249 Chr.5: 96879810-96879810 on Build GRCh38
rs770441171 Chr.5: 96879890-96879890 on Build GRCh38
rs758416048 Chr.5: 96879812-96879812 on Build GRCh38
rs776367183 Chr.5: 96879794-96879794 on Build GRCh38
rs764098841 Chr.5: 96879816-96879816 on Build GRCh38
rs753850576 Chr.5: 96879966-96879966 on Build GRCh38
rs61731315 Chr.5: 96879878-96879878 on Build GRCh38
rs376529950 Chr.5: 96879926-96879926 on Build GRCh38
rs749534634 Chr.5: 96879848-96879848 on Build GRCh38
rs755606139 Chr.5: 96879891-96879892 on Build GRCh38
rs775400779 Chr.5: 96879921-96879921 on Build GRCh38
rs758783383 Chr.5: 96879974-96879974 on Build GRCh38
rs755150805 Chr.5: 96879860-96879860 on Build GRCh38
rs369951421 Chr.5: 96879973-96879973 on Build GRCh38
rs140447588 Chr.5: 96879781-96879781 on Build GRCh38
rs772760308 Chr.5: 96879782-96879782 on Build GRCh38
rs1030038805 Chr.5: 96879809-96879809 on Build GRCh38
rs765833440 Chr.5: 96879790-96879790 on Build GRCh38
rs751505544 Chr.5: 96879819-96879819 on Build GRCh38
rs560867442 Chr.5: 96879970-96879970 on Build GRCh38
rs202185665 Chr.5: 96879889-96879889 on Build GRCh38
rs760144825 Chr.5: 96879789-96879789 on Build GRCh38
rs199924315 Chr.5: 96879870-96879870 on Build GRCh38
rs773737033 Chr.5: 96879887-96879887 on Build GRCh38
rs369000300 Chr.5: 96879960-96879960 on Build GRCh38
rs780227787 Chr.5: 96879833-96879833 on Build GRCh38
rs756287293 Chr.5: 96879822-96879822 on Build GRCh38
rs41506651 Chr.5: 96879976-96879976 on Build GRCh38
rs544718303 Chr.5: 96879892-96879892 on Build GRCh38
rs190219466 Chr.5: 96879939-96879939 on Build GRCh38
rs759295580 Chr.5: 96879898-96879898 on Build GRCh38
rs759219335 Chr.5: 96879832-96879832 on Build GRCh38
rs747518575 Chr.5: 96879982-96879982 on Build GRCh38
rs530358131 Chr.5: 96879922-96879922 on Build GRCh38
rs765826326 Chr.5: 96879931-96879931 on Build GRCh38
rs1027563963 Chr.5: 96879962-96879962 on Build GRCh38
rs778973997 Chr.5: 96879971-96879971 on Build GRCh38
rs757198350 Chr.5: 96879934-96879934 on Build GRCh38
rs868684737 Chr.5: 96879905-96879905 on Build GRCh38
rs371722634 Chr.5: 96879904-96879904 on Build GRCh38
rs765081459 Chr.5: 96879801-96879801 on Build GRCh38
rs763935468 Chr.5: 96879924-96879924 on Build GRCh38
rs367736564 Chr.5: 96879777-96879777 on Build GRCh38
rs527993385 Chr.5: 96879975-96879975 on Build GRCh38
rs565149554 Chr.5: 96879867-96879867 on Build GRCh38
rs748758410 Chr.5: 96879779-96879779 on Build GRCh38
rs945074898 Chr.5: 96879958-96879959 on Build GRCh38
rs375600819 Chr.5: 96879861-96879861 on Build GRCh38
rs752046082 Chr.5: 96879887-96879895 on Build GRCh38
rs763467396 Chr.5: 96879841-96879841 on Build GRCh38
rs779392672 Chr.5: 96879866-96879866 on Build GRCh38

View Full Product Details