Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73447379-73447606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFP
Entrez Gene ID
174
Gene Name
alpha fetoprotein
Gene Aliases
AFPD, FETA, HPAFP
UniGene
Hs.518808
Gene Chromosome Location
Chr.4: 73436219-73455785 on Build GRCh38
Transcripts
NM_001134.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4660257 c.929G>A p.R310H Chr.4: 73447547-73447547 on Build GRCh38
3605693 c.980C>T p.S327F Chr.4: 73447598-73447598 on Build GRCh38
5452146 c.845A>G p.E282G Chr.4: 73447463-73447463 on Build GRCh38
4827392 c.936A>T p.Q312H Chr.4: 73447554-73447554 on Build GRCh38
5931442 c.844G>A p.E282K Chr.4: 73447462-73447462 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745393842 Chr.4: 73447574-73447576 on Build GRCh38
rs1034676137 Chr.4: 73447474-73447474 on Build GRCh38
rs1043658246 Chr.4: 73447567-73447567 on Build GRCh38
rs566738802 Chr.4: 73447518-73447518 on Build GRCh38
rs995789230 Chr.4: 73447471-73447471 on Build GRCh38
rs967861911 Chr.4: 73447559-73447559 on Build GRCh38
rs747788767 Chr.4: 73447470-73447470 on Build GRCh38
rs369341269 Chr.4: 73447511-73447511 on Build GRCh38
rs755337108 Chr.4: 73447453-73447453 on Build GRCh38
rs757216268 Chr.4: 73447417-73447417 on Build GRCh38
rs387906580 Chr.4: 73447501-73447502 on Build GRCh38
rs147848862 Chr.4: 73447503-73447503 on Build GRCh38
rs931904529 Chr.4: 73447555-73447555 on Build GRCh38
rs778772992 Chr.4: 73447462-73447463 on Build GRCh38
rs760761382 Chr.4: 73447538-73447538 on Build GRCh38
rs766531416 Chr.4: 73447417-73447417 on Build GRCh38
rs1027150427 Chr.4: 73447475-73447475 on Build GRCh38
rs1003646245 Chr.4: 73447473-73447473 on Build GRCh38
rs773339360 Chr.4: 73447537-73447537 on Build GRCh38
rs762166115 Chr.4: 73447420-73447420 on Build GRCh38
rs7667494 Chr.4: 73447452-73447452 on Build GRCh38
rs966201909 Chr.4: 73447521-73447521 on Build GRCh38
rs753006750 Chr.4: 73447554-73447554 on Build GRCh38
rs756549744 Chr.4: 73447437-73447437 on Build GRCh38
rs776490501 Chr.4: 73447493-73447493 on Build GRCh38
rs765025434 Chr.4: 73447412-73447412 on Build GRCh38
rs925113628 Chr.4: 73447569-73447569 on Build GRCh38
rs760178436 Chr.4: 73447540-73447540 on Build GRCh38
rs746485734 Chr.4: 73447492-73447492 on Build GRCh38
rs768059598 Chr.4: 73447539-73447539 on Build GRCh38
rs28482344 Chr.4: 73447476-73447476 on Build GRCh38
rs765672473 Chr.4: 73447546-73447546 on Build GRCh38
rs767485566 Chr.4: 73447426-73447426 on Build GRCh38
rs758117253 Chr.4: 73447472-73447472 on Build GRCh38
rs779486958 Chr.4: 73447466-73447466 on Build GRCh38
rs774494158 Chr.4: 73447414-73447414 on Build GRCh38
rs761922000 Chr.4: 73447519-73447519 on Build GRCh38
rs144523159 Chr.4: 73447499-73447499 on Build GRCh38
rs867508994 Chr.4: 73447526-73447526 on Build GRCh38
rs767824349 Chr.4: 73447536-73447536 on Build GRCh38
rs766870141 Chr.4: 73447449-73447449 on Build GRCh38
rs750566810 Chr.4: 73447427-73447427 on Build GRCh38

View Full Product Details