Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 74291816-74292053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5A
Entrez Gene ID
1528
Gene Name
cytochrome b5 type A
Gene Aliases
CYB5, MCB5
UniGene
Hs.465413
Gene Chromosome Location
Chr.18: 74253292-74292016 on Build GRCh38
Transcripts
NM_001190807.2 NM_001914.3 NM_148923.3 XM_011525835.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
280245 c.48G>T p.E16D Chr.18: 74291828-74291828 on Build GRCh38
438375 c.45G>A p.E15E Chr.18: 74291831-74291831 on Build GRCh38
4073214 c.22G>A p.A8T Chr.18: 74291854-74291854 on Build GRCh38
3146219 c.31T>C p.Y11H Chr.18: 74291845-74291845 on Build GRCh38
3146220 c.7G>C p.E3Q Chr.18: 74291869-74291869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547120179 Chr.18: 74291918-74291918 on Build GRCh38
rs1051236 Chr.18: 74291840-74291840 on Build GRCh38
rs770210317 Chr.18: 74291896-74291896 on Build GRCh38
rs201724841 Chr.18: 74291922-74291922 on Build GRCh38
rs372498253 Chr.18: 74291883-74291883 on Build GRCh38
rs943034746 Chr.18: 74291953-74291953 on Build GRCh38
rs550423813 Chr.18: 74292007-74292007 on Build GRCh38
rs878970654 Chr.18: 74291956-74291956 on Build GRCh38
rs746210306 Chr.18: 74291888-74291888 on Build GRCh38
rs1041826039 Chr.18: 74292026-74292026 on Build GRCh38
rs201644903 Chr.18: 74291851-74291851 on Build GRCh38
rs529361433 Chr.18: 74291984-74291984 on Build GRCh38
rs907690903 Chr.18: 74291966-74291966 on Build GRCh38
rs560684215 Chr.18: 74291943-74291943 on Build GRCh38
rs766259647 Chr.18: 74291909-74291909 on Build GRCh38
rs756018585 Chr.18: 74291861-74291861 on Build GRCh38
rs536967986 Chr.18: 74291991-74291991 on Build GRCh38
rs533326035 Chr.18: 74291911-74291911 on Build GRCh38
rs750209113 Chr.18: 74291852-74291852 on Build GRCh38
rs768103280 Chr.18: 74291996-74291996 on Build GRCh38
rs371426321 Chr.18: 74291989-74291989 on Build GRCh38
rs981515640 Chr.18: 74291838-74291838 on Build GRCh38
rs952111960 Chr.18: 74291910-74291910 on Build GRCh38
rs901623806 Chr.18: 74291985-74291985 on Build GRCh38
rs200736659 Chr.18: 74291879-74291879 on Build GRCh38
rs780130050 Chr.18: 74291862-74291862 on Build GRCh38
rs754878118 Chr.18: 74291869-74291869 on Build GRCh38
rs944501713 Chr.18: 74291972-74291972 on Build GRCh38
rs531270678 Chr.18: 74291843-74291843 on Build GRCh38
rs777443430 Chr.18: 74291885-74291885 on Build GRCh38
rs956228194 Chr.18: 74291854-74291854 on Build GRCh38
rs75160992 Chr.18: 74291863-74291863 on Build GRCh38
rs771717839 Chr.18: 74291884-74291884 on Build GRCh38
rs548402150 Chr.18: 74291986-74291986 on Build GRCh38
rs763457593 Chr.18: 74291914-74291914 on Build GRCh38
rs369980435 Chr.18: 74291984-74291984 on Build GRCh38
rs1040122670 Chr.18: 74291954-74291954 on Build GRCh38

View Full Product Details