Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 82354484-82354713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC59
Entrez Gene ID
29080
Gene Name
coiled-coil domain containing 59
Gene Aliases
BR22, HSPC128, TAP26
UniGene
Hs.582627
Gene Chromosome Location
Chr.12: 82352304-82358805 on Build GRCh38
Transcripts
NM_014167.4 NR_033192.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5000371 c.561_564+4delACAAGTAA p.? Chr.12: 82354491-82354498 on Build GRCh38
4983407 c.503C>G p.S168* Chr.12: 82354556-82354556 on Build GRCh38
4957312 c.560A>C p.K187T Chr.12: 82354499-82354499 on Build GRCh38
7211215 c.497A>G p.K166R Chr.12: 82354562-82354562 on Build GRCh38
1993764 c.548G>A p.R183H Chr.12: 82354511-82354511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903050015 Chr.12: 82354647-82354647 on Build GRCh38
rs776211244 Chr.12: 82354584-82354584 on Build GRCh38
rs1008441391 Chr.12: 82354683-82354683 on Build GRCh38
rs765709196 Chr.12: 82354593-82354593 on Build GRCh38
rs567392617 Chr.12: 82354648-82354648 on Build GRCh38
rs932431961 Chr.12: 82354652-82354652 on Build GRCh38
rs1049375510 Chr.12: 82354670-82354670 on Build GRCh38
rs866988343 Chr.12: 82354545-82354545 on Build GRCh38
rs758917272 Chr.12: 82354511-82354511 on Build GRCh38
rs775468090 Chr.12: 82354571-82354571 on Build GRCh38
rs138760241 Chr.12: 82354522-82354522 on Build GRCh38
rs1056964118 Chr.12: 82354676-82354676 on Build GRCh38
rs890804565 Chr.12: 82354681-82354681 on Build GRCh38
rs115308705 Chr.12: 82354618-82354618 on Build GRCh38
rs143136847 Chr.12: 82354560-82354560 on Build GRCh38
rs192072744 Chr.12: 82354514-82354514 on Build GRCh38
rs199552914 Chr.12: 82354578-82354578 on Build GRCh38
rs750767488 Chr.12: 82354602-82354602 on Build GRCh38
rs748400737 Chr.12: 82354524-82354524 on Build GRCh38
rs763581240 Chr.12: 82354603-82354603 on Build GRCh38
rs761461434 Chr.12: 82354590-82354590 on Build GRCh38
rs770671883 Chr.12: 82354626-82354626 on Build GRCh38
rs572477012 Chr.12: 82354642-82354642 on Build GRCh38
rs150615186 Chr.12: 82354512-82354512 on Build GRCh38
rs945202701 Chr.12: 82354604-82354604 on Build GRCh38
rs925015125 Chr.12: 82354650-82354650 on Build GRCh38
rs760154289 Chr.12: 82354575-82354575 on Build GRCh38
rs772388515 Chr.12: 82354682-82354688 on Build GRCh38
rs374928926 Chr.12: 82354516-82354516 on Build GRCh38
rs767030736 Chr.12: 82354611-82354611 on Build GRCh38
rs778625853 Chr.12: 82354557-82354557 on Build GRCh38
rs557111104 Chr.12: 82354552-82354552 on Build GRCh38
rs745719520 Chr.12: 82354580-82354580 on Build GRCh38
rs947161885 Chr.12: 82354568-82354568 on Build GRCh38
rs913699582 Chr.12: 82354594-82354594 on Build GRCh38
rs756983003 Chr.12: 82354571-82354571 on Build GRCh38
rs992485919 Chr.12: 82354570-82354570 on Build GRCh38

View Full Product Details