Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32792511-32792772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3B
Entrez Gene ID
1789
Gene Name
DNA methyltransferase 3 beta
Gene Aliases
ICF, ICF1, M.HsaIIIB
UniGene
Hs.643024
Gene Chromosome Location
Chr.20: 32762385-32809356 on Build GRCh38
Transcripts
NM_001207055.1 NM_001207056.1 NM_006892.3 NM_175848.1 NM_175849.1 NM_175850.2 XM_011528653.2 XM_011528654.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1641323 c.1042G>C p.A348P Chr.20: 32792710-32792710 on Build GRCh38
4752115 c.1010A>C p.H337P Chr.20: 32792714-32792714 on Build GRCh38
4678917 c.995T>C p.M332T Chr.20: 32792699-32792699 on Build GRCh38
4678918 c.1031T>C p.M344T Chr.20: 32792699-32792699 on Build GRCh38
6956822 c.996G>A p.M332I Chr.20: 32792700-32792700 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760264212 Chr.20: 32792635-32792635 on Build GRCh38
rs190567237 Chr.20: 32792599-32792599 on Build GRCh38
rs763435705 Chr.20: 32792639-32792639 on Build GRCh38
rs951488082 Chr.20: 32792644-32792644 on Build GRCh38
rs150148922 Chr.20: 32792739-32792739 on Build GRCh38
rs771875972 Chr.20: 32792631-32792631 on Build GRCh38
rs749242142 Chr.20: 32792601-32792601 on Build GRCh38
rs764423053 Chr.20: 32792593-32792593 on Build GRCh38
rs771931437 Chr.20: 32792599-32792600 on Build GRCh38
rs368420286 Chr.20: 32792577-32792577 on Build GRCh38
rs980731277 Chr.20: 32792570-32792570 on Build GRCh38
rs951435928 Chr.20: 32792533-32792533 on Build GRCh38
rs886056613 Chr.20: 32792664-32792664 on Build GRCh38
rs773216636 Chr.20: 32792716-32792716 on Build GRCh38
rs778738485 Chr.20: 32792592-32792593 on Build GRCh38
rs747983703 Chr.20: 32792725-32792725 on Build GRCh38
rs747782702 Chr.20: 32792628-32792628 on Build GRCh38
rs970776221 Chr.20: 32792715-32792715 on Build GRCh38
rs761984399 Chr.20: 32792659-32792659 on Build GRCh38
rs767733477 Chr.20: 32792666-32792666 on Build GRCh38
rs757945003 Chr.20: 32792665-32792665 on Build GRCh38
rs764487838 Chr.20: 32792645-32792645 on Build GRCh38
rs140978291 Chr.20: 32792677-32792677 on Build GRCh38
rs758800641 Chr.20: 32792581-32792581 on Build GRCh38
rs757508182 Chr.20: 32792597-32792597 on Build GRCh38
rs759862311 Chr.20: 32792712-32792712 on Build GRCh38
rs768497451 Chr.20: 32792612-32792612 on Build GRCh38
rs778384740 Chr.20: 32792566-32792566 on Build GRCh38
rs375596097 Chr.20: 32792598-32792598 on Build GRCh38
rs865883633 Chr.20: 32792689-32792689 on Build GRCh38
rs368635669 Chr.20: 32792681-32792681 on Build GRCh38
rs777499803 Chr.20: 32792669-32792669 on Build GRCh38
rs375043948 Chr.20: 32792576-32792576 on Build GRCh38
rs752933235 Chr.20: 32792579-32792579 on Build GRCh38
rs772806134 Chr.20: 32792634-32792634 on Build GRCh38
rs776048512 Chr.20: 32792637-32792637 on Build GRCh38
rs780068965 Chr.20: 32792679-32792679 on Build GRCh38
rs982820224 Chr.20: 32792559-32792559 on Build GRCh38
rs975587740 Chr.20: 32792594-32792594 on Build GRCh38
rs1013448701 Chr.20: 32792716-32792716 on Build GRCh38
rs754921958 Chr.20: 32792720-32792720 on Build GRCh38
rs138369083 Chr.20: 32792651-32792651 on Build GRCh38
rs770469129 Chr.20: 32792744-32792744 on Build GRCh38
rs949384697 Chr.20: 32792568-32792568 on Build GRCh38
rs771781648 Chr.20: 32792731-32792731 on Build GRCh38
rs6088006 Chr.20: 32792541-32792541 on Build GRCh38
rs748074327 Chr.20: 32792593-32792593 on Build GRCh38
rs995710171 Chr.20: 32792617-32792617 on Build GRCh38
rs186977189 Chr.20: 32792596-32792596 on Build GRCh38
rs200508870 Chr.20: 32792618-32792618 on Build GRCh38

View Full Product Details