Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107758751-107759022 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLL1
Entrez Gene ID
79872
Gene Name
Cbl proto-oncogene like 1
Gene Aliases
HAKAI, RNF188
UniGene
Hs.592271
Gene Chromosome Location
Chr.7: 107743497-107764997 on Build GRCh38
Transcripts
NM_001284291.1 NM_024814.3 NR_024199.2 XM_011516580.2 XM_017012643.1 XM_017012644.1 XM_017012645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
23447 c.1119A>G p.P373P Chr.7: 107758821-107758821 on Build GRCh38
3631793 c.1291C>T p.Q431* Chr.7: 107758993-107758993 on Build GRCh38
7250171 c.1141C>G p.P381A Chr.7: 107758843-107758843 on Build GRCh38
6175714 c.1317G>C p.L439L Chr.7: 107759019-107759019 on Build GRCh38
1724484 c.1137_1138delTG p.A380fs*45 Chr.7: 107758839-107758840 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753214879 Chr.7: 107758786-107758786 on Build GRCh38
rs370363717 Chr.7: 107758925-107758925 on Build GRCh38
rs992840878 Chr.7: 107758991-107758991 on Build GRCh38
rs1002411012 Chr.7: 107758895-107758895 on Build GRCh38
rs757646034 Chr.7: 107758820-107758820 on Build GRCh38
rs746971001 Chr.7: 107758896-107758896 on Build GRCh38
rs768550180 Chr.7: 107758904-107758904 on Build GRCh38
rs780170009 Chr.7: 107758892-107758892 on Build GRCh38
rs377697227 Chr.7: 107758923-107758923 on Build GRCh38
rs764716196 Chr.7: 107758961-107758961 on Build GRCh38
rs141234740 Chr.7: 107758970-107758970 on Build GRCh38
rs148276066 Chr.7: 107758821-107758821 on Build GRCh38
rs765850580 Chr.7: 107758777-107758777 on Build GRCh38
rs747963861 Chr.7: 107758913-107758913 on Build GRCh38
rs757700907 Chr.7: 107758974-107758974 on Build GRCh38
rs150861689 Chr.7: 107758798-107758798 on Build GRCh38
rs113077223 Chr.7: 107758799-107758799 on Build GRCh38
rs765853157 Chr.7: 107758948-107758948 on Build GRCh38
rs756618872 Chr.7: 107758792-107758792 on Build GRCh38
rs887249765 Chr.7: 107758877-107758877 on Build GRCh38
rs765690554 Chr.7: 107758992-107758992 on Build GRCh38
rs746033563 Chr.7: 107758844-107758844 on Build GRCh38
rs764591252 Chr.7: 107758808-107758808 on Build GRCh38
rs754281764 Chr.7: 107758815-107758815 on Build GRCh38
rs773879228 Chr.7: 107758951-107758951 on Build GRCh38
rs758472832 Chr.7: 107758863-107758863 on Build GRCh38
rs199798345 Chr.7: 107758959-107758959 on Build GRCh38
rs765583287 Chr.7: 107758946-107758946 on Build GRCh38
rs904594866 Chr.7: 107758917-107758917 on Build GRCh38

View Full Product Details