Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 86158848-86159107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGBL1
Entrez Gene ID
123624
Gene Name
ATP/GTP binding protein like 1
Gene Aliases
CCP4, FECD8
UniGene
Hs.31795
Gene Chromosome Location
Chr.15: 86079635-87031476 on Build GRCh38
Transcripts
XM_011521226.2 XM_011521227.2 XM_011521228.2 XM_011521230.2 XM_017021918.1 XM_017021919.1 XM_017021920.1 XM_017021921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6522341 c.344C>A p.A115E Chr.15: 86159020-86159020 on Build GRCh38
7352460 c.334C>T p.R112C Chr.15: 86159010-86159010 on Build GRCh38
4616754 c.258C>A p.V86V Chr.15: 86158934-86158934 on Build GRCh38
5923105 c.277G>A p.G93R Chr.15: 86158953-86158953 on Build GRCh38
6143262 c.310G>C p.V104L Chr.15: 86158986-86158986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780777593 Chr.15: 86158922-86158922 on Build GRCh38
rs376965409 Chr.15: 86159045-86159045 on Build GRCh38
rs753586551 Chr.15: 86159069-86159069 on Build GRCh38
rs772534565 Chr.15: 86158996-86158996 on Build GRCh38
rs763940129 Chr.15: 86158909-86158909 on Build GRCh38
rs755789582 Chr.15: 86158945-86158945 on Build GRCh38
rs748755624 Chr.15: 86158956-86158956 on Build GRCh38
rs761775810 Chr.15: 86159039-86159039 on Build GRCh38
rs572239525 Chr.15: 86158938-86158938 on Build GRCh38
rs79814209 Chr.15: 86159010-86159010 on Build GRCh38
rs201712819 Chr.15: 86159004-86159004 on Build GRCh38
rs769734136 Chr.15: 86159065-86159066 on Build GRCh38
rs766442635 Chr.15: 86158895-86158895 on Build GRCh38
rs778607249 Chr.15: 86159076-86159076 on Build GRCh38
rs767255493 Chr.15: 86159043-86159043 on Build GRCh38
rs747867637 Chr.15: 86159029-86159030 on Build GRCh38
rs145790658 Chr.15: 86158971-86158971 on Build GRCh38
rs553816742 Chr.15: 86158877-86158877 on Build GRCh38
rs375766500 Chr.15: 86158890-86158890 on Build GRCh38
rs369571083 Chr.15: 86159005-86159005 on Build GRCh38
rs571192563 Chr.15: 86158907-86158907 on Build GRCh38
rs755728559 Chr.15: 86159052-86159052 on Build GRCh38
rs765414052 Chr.15: 86159021-86159021 on Build GRCh38
rs775341691 Chr.15: 86159011-86159011 on Build GRCh38
rs751561654 Chr.15: 86158936-86158936 on Build GRCh38
rs546354844 Chr.15: 86158947-86158947 on Build GRCh38
rs759528910 Chr.15: 86159014-86159014 on Build GRCh38
rs768281107 Chr.15: 86158968-86158968 on Build GRCh38
rs762888475 Chr.15: 86158906-86158906 on Build GRCh38
rs917111125 Chr.15: 86158952-86158952 on Build GRCh38
rs754425920 Chr.15: 86159073-86159073 on Build GRCh38
rs753885400 Chr.15: 86158902-86158902 on Build GRCh38
rs1029741547 Chr.15: 86159032-86159032 on Build GRCh38
rs929803833 Chr.15: 86158976-86158976 on Build GRCh38
rs1022560369 Chr.15: 86159048-86159048 on Build GRCh38
rs1009019062 Chr.15: 86158916-86158916 on Build GRCh38
rs531237010 Chr.15: 86159034-86159034 on Build GRCh38
rs191671121 Chr.15: 86159053-86159053 on Build GRCh38
rs369256384 Chr.15: 86158944-86158944 on Build GRCh38

View Full Product Details