Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54145677-54145902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT3
Entrez Gene ID
4849
Gene Name
CCR4-NOT transcription complex subunit 3
Gene Aliases
LENG2, NOT3, NOT3H
UniGene
Hs.343571
Gene Chromosome Location
Chr.19: 54137689-54155708 on Build GRCh38
Transcripts
NM_014516.3 XM_005278279.1 XM_005278280.2 XM_005278281.1 XM_005278282.2 XM_011526992.1 XM_011526993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5751983 c.160+1G>AG p.? Chr.19: 54145818-54145818 on Build GRCh38
1612766 c.691C>T p.L231F Chr.19: 54145805-54145805 on Build GRCh38
3835668 c.102G>T p.Q34H Chr.19: 54145759-54145759 on Build GRCh38
5818233 c.606G>T p.K202N Chr.19: 54145720-54145720 on Build GRCh38
4081189 c.652G>A p.D218N Chr.19: 54145766-54145766 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199874706 Chr.19: 54145856-54145856 on Build GRCh38
rs773254399 Chr.19: 54145855-54145855 on Build GRCh38
rs375211815 Chr.19: 54145863-54145863 on Build GRCh38
rs372705849 Chr.19: 54145810-54145810 on Build GRCh38
rs747901070 Chr.19: 54145853-54145853 on Build GRCh38
rs765407340 Chr.19: 54145874-54145874 on Build GRCh38
rs748810728 Chr.19: 54145780-54145780 on Build GRCh38
rs375948136 Chr.19: 54145817-54145817 on Build GRCh38
rs779539051 Chr.19: 54145703-54145703 on Build GRCh38
rs775714798 Chr.19: 54145878-54145878 on Build GRCh38
rs34286651 Chr.19: 54145702-54145702 on Build GRCh38
rs144796513 Chr.19: 54145708-54145708 on Build GRCh38
rs141264166 Chr.19: 54145792-54145792 on Build GRCh38
rs759861030 Chr.19: 54145793-54145793 on Build GRCh38
rs776719450 Chr.19: 54145866-54145866 on Build GRCh38
rs772043379 Chr.19: 54145854-54145854 on Build GRCh38
rs191843066 Chr.19: 54145825-54145825 on Build GRCh38
rs774229944 Chr.19: 54145733-54145733 on Build GRCh38
rs73936620 Chr.19: 54145835-54145835 on Build GRCh38
rs34936413 Chr.19: 54145765-54145765 on Build GRCh38
rs952488775 Chr.19: 54145720-54145720 on Build GRCh38
rs373852003 Chr.19: 54145841-54145841 on Build GRCh38
rs369057695 Chr.19: 54145838-54145838 on Build GRCh38
rs770901969 Chr.19: 54145862-54145862 on Build GRCh38
rs369073653 Chr.19: 54145867-54145867 on Build GRCh38
rs755778475 Chr.19: 54145865-54145865 on Build GRCh38
rs772094308 Chr.19: 54145746-54145746 on Build GRCh38
rs35511193 Chr.19: 54145771-54145771 on Build GRCh38
rs587608163 Chr.19: 54145729-54145729 on Build GRCh38
rs112551303 Chr.19: 54145833-54145833 on Build GRCh38
rs765395773 Chr.19: 54145807-54145807 on Build GRCh38
rs765894239 Chr.19: 54145850-54145850 on Build GRCh38
rs778738017 Chr.19: 54145845-54145845 on Build GRCh38
rs367791191 Chr.19: 54145744-54145744 on Build GRCh38
rs921159903 Chr.19: 54145823-54145823 on Build GRCh38
rs879222513 Chr.19: 54145869-54145869 on Build GRCh38

View Full Product Details