Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6741680-6741917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AGPAT5
Entrez Gene ID
55326
Gene Name
1-acylglycerol-3-phosphate O-acyltransferase 5
Gene Aliases
1AGPAT5, LPAATE
UniGene
Hs.624002
Gene Chromosome Location
Chr.8: 6708357-6761503 on Build GRCh38
Transcripts
NM_018361.3 XM_011534751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5628154 c.575C>T p.A192V Chr.8: 6741740-6741740 on Build GRCh38
6596398 c.523A>G p.T175A Chr.8: 6741688-6741688 on Build GRCh38
308992 c.574G>A p.A192T Chr.8: 6741739-6741739 on Build GRCh38
1686569 c.586+1G>T p.? Chr.8: 6741752-6741752 on Build GRCh38
3650180 c.535C>T p.P179S Chr.8: 6741700-6741700 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs142454856 Chr.8: 6741748-6741748 on Build GRCh38
rs765360549 Chr.8: 6741741-6741741 on Build GRCh38
rs779755430 Chr.8: 6741800-6741800 on Build GRCh38
rs983444070 Chr.8: 6741812-6741812 on Build GRCh38
rs751337545 Chr.8: 6741776-6741776 on Build GRCh38
rs751588800 Chr.8: 6741746-6741747 on Build GRCh38
rs200006372 Chr.8: 6741777-6741777 on Build GRCh38
rs376651920 Chr.8: 6741726-6741726 on Build GRCh38
rs780006107 Chr.8: 6741762-6741763 on Build GRCh38
rs142001193 Chr.8: 6741710-6741710 on Build GRCh38
rs780998745 Chr.8: 6741788-6741788 on Build GRCh38
rs932144894 Chr.8: 6741882-6741882 on Build GRCh38
rs199671502 Chr.8: 6741785-6741785 on Build GRCh38
rs756879482 Chr.8: 6741782-6741782 on Build GRCh38
rs1021492034 Chr.8: 6741708-6741708 on Build GRCh38
rs183702643 Chr.8: 6741804-6741804 on Build GRCh38
rs537592148 Chr.8: 6741806-6741806 on Build GRCh38
rs766609307 Chr.8: 6741731-6741731 on Build GRCh38
rs752609216 Chr.8: 6741742-6741742 on Build GRCh38
rs973454198 Chr.8: 6741858-6741858 on Build GRCh38
rs963471539 Chr.8: 6741834-6741834 on Build GRCh38
rs200744401 Chr.8: 6741829-6741829 on Build GRCh38
rs200547380 Chr.8: 6741712-6741712 on Build GRCh38
rs755668602 Chr.8: 6741792-6741792 on Build GRCh38
rs756076142 Chr.8: 6741755-6741766 on Build GRCh38
rs749199581 Chr.8: 6741802-6741802 on Build GRCh38
rs764090973 Chr.8: 6741744-6741745 on Build GRCh38
rs772486814 Chr.8: 6741803-6741803 on Build GRCh38
rs772455784 Chr.8: 6741721-6741721 on Build GRCh38
rs753855805 Chr.8: 6741732-6741732 on Build GRCh38
rs367679815 Chr.8: 6741765-6741765 on Build GRCh38
rs922008351 Chr.8: 6741879-6741879 on Build GRCh38
rs372688517 Chr.8: 6741718-6741718 on Build GRCh38
rs111800074 Chr.8: 6741813-6741813 on Build GRCh38
rs151313617 Chr.8: 6741749-6741749 on Build GRCh38
rs759609418 Chr.8: 6741740-6741740 on Build GRCh38
rs774992197 Chr.8: 6741717-6741717 on Build GRCh38
rs907864303 Chr.8: 6741827-6741827 on Build GRCh38

View Full Product Details