Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55649194-55649457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC106
Entrez Gene ID
29903
Gene Name
coiled-coil domain containing 106
Gene Aliases
HSU79303, ZNF581
UniGene
Hs.82482
Gene Chromosome Location
Chr.19: 55647588-55653161 on Build GRCh38
Transcripts
NM_013301.2 XM_005258827.2 XM_005258828.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7101356 c.52G>C p.E18Q Chr.19: 55649225-55649225 on Build GRCh38
1751171 c.52G>A p.E18K Chr.19: 55649225-55649225 on Build GRCh38
1001582 c.55A>G p.I19V Chr.19: 55649228-55649228 on Build GRCh38
6621249 c.69C>T p.F23F Chr.19: 55649242-55649242 on Build GRCh38
4969538 c.163C>G p.L55V Chr.19: 55649434-55649434 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376647319 Chr.19: 55649386-55649386 on Build GRCh38
rs749076695 Chr.19: 55649289-55649289 on Build GRCh38
rs149619834 Chr.19: 55649321-55649321 on Build GRCh38
rs755654171 Chr.19: 55649358-55649358 on Build GRCh38
rs900776166 Chr.19: 55649323-55649323 on Build GRCh38
rs770674089 Chr.19: 55649329-55649330 on Build GRCh38
rs761171001 Chr.19: 55649230-55649230 on Build GRCh38
rs148314079 Chr.19: 55649435-55649435 on Build GRCh38
rs550542661 Chr.19: 55649388-55649388 on Build GRCh38
rs772213366 Chr.19: 55649305-55649305 on Build GRCh38
rs754826890 Chr.19: 55649381-55649381 on Build GRCh38
rs373494026 Chr.19: 55649412-55649412 on Build GRCh38
rs765941141 Chr.19: 55649430-55649430 on Build GRCh38
rs773296878 Chr.19: 55649320-55649320 on Build GRCh38
rs752965865 Chr.19: 55649331-55649331 on Build GRCh38
rs559216380 Chr.19: 55649225-55649225 on Build GRCh38
rs780135952 Chr.19: 55649288-55649288 on Build GRCh38
rs371601495 Chr.19: 55649295-55649295 on Build GRCh38
rs765656511 Chr.19: 55649329-55649329 on Build GRCh38
rs372401653 Chr.19: 55649385-55649385 on Build GRCh38
rs766817832 Chr.19: 55649233-55649233 on Build GRCh38
rs776524505 Chr.19: 55649402-55649402 on Build GRCh38
rs565915933 Chr.19: 55649272-55649272 on Build GRCh38
rs778706158 Chr.19: 55649436-55649436 on Build GRCh38
rs759781801 Chr.19: 55649382-55649383 on Build GRCh38
rs960412194 Chr.19: 55649394-55649394 on Build GRCh38
rs767264002 Chr.19: 55649352-55649352 on Build GRCh38
rs776447490 Chr.19: 55649331-55649332 on Build GRCh38
rs369658542 Chr.19: 55649398-55649398 on Build GRCh38
rs760120672 Chr.19: 55649404-55649404 on Build GRCh38
rs201095990 Chr.19: 55649351-55649351 on Build GRCh38
rs551680180 Chr.19: 55649286-55649286 on Build GRCh38
rs199848148 Chr.19: 55649410-55649410 on Build GRCh38
rs748431578 Chr.19: 55649298-55649298 on Build GRCh38
rs751204132 Chr.19: 55649269-55649269 on Build GRCh38
rs777167908 Chr.19: 55649325-55649325 on Build GRCh38
rs750078376 Chr.19: 55649423-55649423 on Build GRCh38
rs201746252 Chr.19: 55649328-55649328 on Build GRCh38
rs529274073 Chr.19: 55649369-55649369 on Build GRCh38
rs770135153 Chr.19: 55649406-55649406 on Build GRCh38
rs763698016 Chr.19: 55649266-55649266 on Build GRCh38
rs141190085 Chr.19: 55649426-55649426 on Build GRCh38
rs763404034 Chr.19: 55649415-55649415 on Build GRCh38
rs868123331 Chr.19: 55649232-55649232 on Build GRCh38
rs367901327 Chr.19: 55649243-55649243 on Build GRCh38
rs927251453 Chr.19: 55649253-55649253 on Build GRCh38
rs920670122 Chr.19: 55649276-55649276 on Build GRCh38
rs148385298 Chr.19: 55649431-55649431 on Build GRCh38
rs761844514 Chr.19: 55649218-55649218 on Build GRCh38
rs114465612 Chr.19: 55649275-55649275 on Build GRCh38
rs771498342 Chr.19: 55649322-55649322 on Build GRCh38
rs754217737 Chr.19: 55649242-55649242 on Build GRCh38
rs201897477 Chr.19: 55649294-55649294 on Build GRCh38
rs773231620 Chr.19: 55649228-55649228 on Build GRCh38
rs767316961 Chr.19: 55649419-55649419 on Build GRCh38

View Full Product Details