Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 85721784-85722057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP24
Entrez Gene ID
83478
Gene Name
Rho GTPase activating protein 24
Gene Aliases
FILGAP, RC-GAP72, RCGAP72, p73, p73RhoGAP
UniGene
Hs.444229
Gene Chromosome Location
Chr.4: 85475131-86002670 on Build GRCh38
Transcripts
NM_001025616.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4921903 c.245A>T p.K82M Chr.4: 85721949-85721949 on Build GRCh38
301691 c.222C>A p.P74P Chr.4: 85721926-85721926 on Build GRCh38
3606750 c.266C>T p.P89L Chr.4: 85721970-85721970 on Build GRCh38
1058409 c.212C>A p.S71Y Chr.4: 85721916-85721916 on Build GRCh38
243022 c.245A>C p.K82T Chr.4: 85721949-85721949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374274489 Chr.4: 85722030-85722030 on Build GRCh38
rs773353957 Chr.4: 85721992-85721992 on Build GRCh38
rs748819927 Chr.4: 85721944-85721944 on Build GRCh38
rs17010697 Chr.4: 85722002-85722002 on Build GRCh38
rs748396958 Chr.4: 85721921-85721921 on Build GRCh38
rs765766682 Chr.4: 85721986-85721986 on Build GRCh38
rs946893722 Chr.4: 85721941-85721941 on Build GRCh38
rs189134282 Chr.4: 85722027-85722027 on Build GRCh38
rs35047755 Chr.4: 85721944-85721945 on Build GRCh38
rs144785317 Chr.4: 85721904-85721904 on Build GRCh38
rs1056520629 Chr.4: 85722000-85722000 on Build GRCh38
rs772099557 Chr.4: 85721923-85721923 on Build GRCh38
rs745658216 Chr.4: 85722023-85722023 on Build GRCh38
rs764632055 Chr.4: 85721844-85721845 on Build GRCh38
rs926703434 Chr.4: 85721990-85721990 on Build GRCh38
rs746810138 Chr.4: 85721919-85721919 on Build GRCh38
rs766678995 Chr.4: 85721978-85721978 on Build GRCh38
rs774930690 Chr.4: 85721943-85721943 on Build GRCh38
rs936017329 Chr.4: 85721817-85721817 on Build GRCh38
rs765281093 Chr.4: 85721880-85721880 on Build GRCh38
rs778038476 Chr.4: 85721915-85721915 on Build GRCh38
rs1055688403 Chr.4: 85721826-85721826 on Build GRCh38
rs575623188 Chr.4: 85721886-85721886 on Build GRCh38
rs768207696 Chr.4: 85721958-85721958 on Build GRCh38
rs145725359 Chr.4: 85721905-85721905 on Build GRCh38
rs530004902 Chr.4: 85722024-85722024 on Build GRCh38
rs184802761 Chr.4: 85722018-85722018 on Build GRCh38
rs200821525 Chr.4: 85721841-85721841 on Build GRCh38
rs745852491 Chr.4: 85721924-85721924 on Build GRCh38
rs912226976 Chr.4: 85721830-85721830 on Build GRCh38
rs769743219 Chr.4: 85721940-85721940 on Build GRCh38
rs755072531 Chr.4: 85721875-85721875 on Build GRCh38
rs754302572 Chr.4: 85721870-85721870 on Build GRCh38
rs761050625 Chr.4: 85721838-85721841 on Build GRCh38
rs761058241 Chr.4: 85721969-85721969 on Build GRCh38
rs6816135 Chr.4: 85721846-85721846 on Build GRCh38
rs1043861824 Chr.4: 85721953-85721953 on Build GRCh38
rs564276651 Chr.4: 85721981-85721981 on Build GRCh38
rs199873264 Chr.4: 85722019-85722019 on Build GRCh38
rs367769099 Chr.4: 85722005-85722005 on Build GRCh38
rs751786204 Chr.4: 85722012-85722012 on Build GRCh38

View Full Product Details