Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 100774382-100774649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI3BP
Entrez Gene ID
25890
Gene Name
ABI family member 3 binding protein
Gene Aliases
NESHBP, TARSH
UniGene
Hs.477015
Gene Chromosome Location
Chr.3: 100749335-100993508 on Build GRCh38
Transcripts
NM_015429.3 XM_005247281.2 XM_005247282.2 XM_005247283.2 XM_005247284.2 XM_005247285.2 XM_005247287.2 XM_005247288.2 XM_005247289.2 XM_005247290.2 XM_005247291.2 XM_005247293.2 XM_005247294.2 XM_005247295.2 XM_005247296.2 XM_005247298.2 XM_005247299.3 XM_005247300.2 XM_005247301.2 XM_005247302.2 XM_005247303.2 XM_005247305.2 XM_005247306.2 XM_005247309.1 XM_006713569.2 XM_006713570.2 XM_006713571.2 XM_006713572.2 XM_011512639.1 XM_011512640.1 XM_011512641.1 XM_011512643.1 XM_011512644.1 XM_011512645.1 XM_011512646.1 XM_011512647.1 XM_011512648.1 XM_011512649.1 XM_011512651.1 XM_011512652.1 XM_011512653.1 XM_011512654.1 XM_011512656.1 XM_011512657.1 XM_011512658.1 XM_011512659.1 XM_011512660.1 XM_017006105.1 XM_017006106.1 XM_017006107.1 XM_017006108.1

SNP Detail

SNP ID Chromosome Location
rs534055674 Chr.3: 100774551-100774551 on Build GRCh38
rs746773188 Chr.3: 100774569-100774569 on Build GRCh38
rs776433521 Chr.3: 100774579-100774579 on Build GRCh38
rs551331501 Chr.3: 100774434-100774434 on Build GRCh38
rs933022380 Chr.3: 100774508-100774509 on Build GRCh38
rs987160215 Chr.3: 100774520-100774520 on Build GRCh38
rs1032196337 Chr.3: 100774447-100774447 on Build GRCh38
rs777762089 Chr.3: 100774566-100774566 on Build GRCh38
rs548772401 Chr.3: 100774564-100774564 on Build GRCh38
rs968512138 Chr.3: 100774593-100774593 on Build GRCh38
rs9846590 Chr.3: 100774578-100774578 on Build GRCh38
rs753977467 Chr.3: 100774423-100774423 on Build GRCh38
rs769409176 Chr.3: 100774583-100774583 on Build GRCh38
rs900731887 Chr.3: 100774445-100774445 on Build GRCh38
rs921590654 Chr.3: 100774502-100774502 on Build GRCh38
rs979873976 Chr.3: 100774604-100774604 on Build GRCh38
rs1034245770 Chr.3: 100774615-100774615 on Build GRCh38
rs997520551 Chr.3: 100774454-100774454 on Build GRCh38
rs1010735344 Chr.3: 100774490-100774490 on Build GRCh38
rs1037141145 Chr.3: 100774546-100774546 on Build GRCh38
rs1025441003 Chr.3: 100774433-100774433 on Build GRCh38
rs774848690 Chr.3: 100774598-100774598 on Build GRCh38
rs537637826 Chr.3: 100774580-100774580 on Build GRCh38
rs940466228 Chr.3: 100774537-100774537 on Build GRCh38
rs957924153 Chr.3: 100774466-100774466 on Build GRCh38
rs907585158 Chr.3: 100774522-100774523 on Build GRCh38
rs116407858 Chr.3: 100774474-100774474 on Build GRCh38
rs891914215 Chr.3: 100774477-100774477 on Build GRCh38

View Full Product Details