Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128262055-128262326 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEFSEC
Entrez Gene ID
60678
Gene Name
eukaryotic elongation factor, selenocysteine-tRNA specific
Gene Aliases
EFSEC, SELB
UniGene
Hs.477498
Gene Chromosome Location
Chr.3: 128153456-128426193 on Build GRCh38
Transcripts
NM_021937.4 XM_005247696.2 XM_017006975.1 XM_017006976.1 XM_017006977.1 XM_017006978.1 XM_017006979.1 XM_017006980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7079791 c.589G>A p.E197K Chr.3: 128262192-128262192 on Build GRCh38
5978717 c.554C>T p.A185V Chr.3: 128262157-128262157 on Build GRCh38
1418750 c.550G>A p.V184M Chr.3: 128262153-128262153 on Build GRCh38
5974530 c.607C>T p.P203S Chr.3: 128262210-128262210 on Build GRCh38
3992709 c.577G>C p.A193P Chr.3: 128262180-128262180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932731005 Chr.3: 128262288-128262288 on Build GRCh38
rs752925399 Chr.3: 128262091-128262091 on Build GRCh38
rs765545914 Chr.3: 128262081-128262081 on Build GRCh38
rs374137398 Chr.3: 128262117-128262117 on Build GRCh38
rs112744437 Chr.3: 128262167-128262167 on Build GRCh38
rs1052407806 Chr.3: 128262296-128262296 on Build GRCh38
rs528203343 Chr.3: 128262186-128262186 on Build GRCh38
rs754774089 Chr.3: 128262259-128262259 on Build GRCh38
rs780105565 Chr.3: 128262239-128262239 on Build GRCh38
rs761378874 Chr.3: 128262144-128262144 on Build GRCh38
rs769591033 Chr.3: 128262135-128262135 on Build GRCh38
rs1050286006 Chr.3: 128262291-128262291 on Build GRCh38
rs764296859 Chr.3: 128262166-128262166 on Build GRCh38
rs551268095 Chr.3: 128262274-128262274 on Build GRCh38
rs750307785 Chr.3: 128262181-128262181 on Build GRCh38
rs370162790 Chr.3: 128262233-128262233 on Build GRCh38
rs377582079 Chr.3: 128262157-128262157 on Build GRCh38
rs61740550 Chr.3: 128262152-128262152 on Build GRCh38
rs780092371 Chr.3: 128262185-128262185 on Build GRCh38
rs546855056 Chr.3: 128262195-128262195 on Build GRCh38
rs756015707 Chr.3: 128262183-128262183 on Build GRCh38
rs569230091 Chr.3: 128262293-128262293 on Build GRCh38
rs770190494 Chr.3: 128262213-128262213 on Build GRCh38
rs746499017 Chr.3: 128262268-128262268 on Build GRCh38
rs568685973 Chr.3: 128262220-128262220 on Build GRCh38
rs771533469 Chr.3: 128262150-128262150 on Build GRCh38
rs991650447 Chr.3: 128262168-128262168 on Build GRCh38
rs922217870 Chr.3: 128262224-128262224 on Build GRCh38
rs758000287 Chr.3: 128262265-128262265 on Build GRCh38
rs781165633 Chr.3: 128262115-128262115 on Build GRCh38
rs542147902 Chr.3: 128262096-128262096 on Build GRCh38
rs138191450 Chr.3: 128262099-128262099 on Build GRCh38
rs7625074 Chr.3: 128262158-128262158 on Build GRCh38
rs377382186 Chr.3: 128262120-128262120 on Build GRCh38
rs776045813 Chr.3: 128262214-128262214 on Build GRCh38
rs187895280 Chr.3: 128262264-128262264 on Build GRCh38
rs767572996 Chr.3: 128262231-128262231 on Build GRCh38
rs373534963 Chr.3: 128262153-128262153 on Build GRCh38
rs953354549 Chr.3: 128262136-128262136 on Build GRCh38
rs753879695 Chr.3: 128262250-128262250 on Build GRCh38
rs768608917 Chr.3: 128262187-128262187 on Build GRCh38
rs373390199 Chr.3: 128262194-128262194 on Build GRCh38
rs781161504 Chr.3: 128262179-128262179 on Build GRCh38
rs116353475 Chr.3: 128262143-128262143 on Build GRCh38
rs917902874 Chr.3: 128262300-128262300 on Build GRCh38
rs760030862 Chr.3: 128262212-128262212 on Build GRCh38
rs777772688 Chr.3: 128262097-128262097 on Build GRCh38
rs781165884 Chr.3: 128262220-128262221 on Build GRCh38
rs747080331 Chr.3: 128262105-128262105 on Build GRCh38
rs965782290 Chr.3: 128262200-128262200 on Build GRCh38
rs369240402 Chr.3: 128262142-128262142 on Build GRCh38
rs779715322 Chr.3: 128262140-128262140 on Build GRCh38
rs771574810 Chr.3: 128262196-128262196 on Build GRCh38
rs916057104 Chr.3: 128262176-128262176 on Build GRCh38
rs761944630 Chr.3: 128262221-128262221 on Build GRCh38
rs757310693 Chr.3: 128262174-128262174 on Build GRCh38
rs772603324 Chr.3: 128262197-128262197 on Build GRCh38
rs757366801 Chr.3: 128262109-128262109 on Build GRCh38

View Full Product Details