Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184317224-184317497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4G1
Entrez Gene ID
1981
Gene Name
eukaryotic translation initiation factor 4 gamma 1
Gene Aliases
EIF-4G1, EIF4F, EIF4G, EIF4GI, P220, PARK18
UniGene
Hs.433750
Gene Chromosome Location
Chr.3: 184314495-184335358 on Build GRCh38
Transcripts
NM_001194946.1 NM_001194947.1 NM_001291157.1 NM_182917.4 NM_198241.2 NM_198242.2 NM_198244.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6855446 c.217C>T p.R73C Chr.3: 184317390-184317390 on Build GRCh38
1617266 c.300delG p.A102fs*63 Chr.3: 184317473-184317473 on Build GRCh38
6306808 c.279G>T p.Q93H Chr.3: 184317452-184317452 on Build GRCh38
446099 c.176C>T p.P59L Chr.3: 184317349-184317349 on Build GRCh38
7118293 c.218G>A p.R73H Chr.3: 184317391-184317391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367958841 Chr.3: 184317340-184317340 on Build GRCh38
rs752230225 Chr.3: 184317345-184317345 on Build GRCh38
rs529763005 Chr.3: 184317275-184317275 on Build GRCh38
rs373769088 Chr.3: 184317371-184317371 on Build GRCh38
rs374124401 Chr.3: 184317296-184317296 on Build GRCh38
rs763266631 Chr.3: 184317308-184317308 on Build GRCh38
rs779901470 Chr.3: 184317390-184317390 on Build GRCh38
rs571849011 Chr.3: 184317349-184317349 on Build GRCh38
rs773258828 Chr.3: 184317297-184317297 on Build GRCh38
rs771346623 Chr.3: 184317426-184317426 on Build GRCh38
rs763590431 Chr.3: 184317441-184317441 on Build GRCh38
rs772052176 Chr.3: 184317283-184317283 on Build GRCh38
rs758297253 Chr.3: 184317385-184317385 on Build GRCh38
rs745750938 Chr.3: 184317282-184317282 on Build GRCh38
rs777911643 Chr.3: 184317411-184317411 on Build GRCh38
rs760027319 Chr.3: 184317444-184317444 on Build GRCh38
rs201980563 Chr.3: 184317393-184317393 on Build GRCh38
rs761253185 Chr.3: 184317477-184317477 on Build GRCh38
rs560273522 Chr.3: 184317264-184317264 on Build GRCh38
rs771352706 Chr.3: 184317332-184317332 on Build GRCh38
rs760113599 Chr.3: 184317407-184317407 on Build GRCh38
rs370719333 Chr.3: 184317295-184317295 on Build GRCh38
rs376639161 Chr.3: 184317472-184317472 on Build GRCh38
rs371421333 Chr.3: 184317341-184317341 on Build GRCh38
rs753368220 Chr.3: 184317356-184317356 on Build GRCh38
rs763694816 Chr.3: 184317351-184317351 on Build GRCh38
rs368386026 Chr.3: 184317285-184317285 on Build GRCh38
rs913697864 Chr.3: 184317360-184317360 on Build GRCh38
rs910334705 Chr.3: 184317389-184317389 on Build GRCh38
rs768180364 Chr.3: 184317343-184317343 on Build GRCh38
rs367604626 Chr.3: 184317455-184317455 on Build GRCh38
rs372316682 Chr.3: 184317461-184317461 on Build GRCh38
rs749410907 Chr.3: 184317420-184317420 on Build GRCh38
rs769721458 Chr.3: 184317396-184317396 on Build GRCh38
rs778662872 Chr.3: 184317373-184317373 on Build GRCh38
rs113810947 Chr.3: 184317384-184317384 on Build GRCh38
rs767148790 Chr.3: 184317274-184317274 on Build GRCh38

View Full Product Details