Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44978246-44978519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B3
Entrez Gene ID
8891
Gene Name
eukaryotic translation initiation factor 2B subunit gamma
Gene Aliases
EIF-2B, EIF2Bgamma
UniGene
Hs.533549
Gene Chromosome Location
Chr.1: 44850522-44986722 on Build GRCh38
Transcripts
NM_001166588.2 NM_001261418.1 NM_020365.4 XM_017002745.1 XM_017002746.1 XM_017002747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
910098 c.191G>A p.C64Y Chr.1: 44978418-44978418 on Build GRCh38
910099 c.185C>T p.A62V Chr.1: 44978424-44978424 on Build GRCh38
681189 c.159G>T p.V53V Chr.1: 44978450-44978450 on Build GRCh38
6039748 c.210_211insA p.M71fs*10 Chr.1: 44978398-44978399 on Build GRCh38
4472546 c.178C>T p.Q60* Chr.1: 44978431-44978431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746995968 Chr.1: 44978355-44978355 on Build GRCh38
rs11556200 Chr.1: 44978366-44978366 on Build GRCh38
rs746090884 Chr.1: 44978334-44978334 on Build GRCh38
rs768258901 Chr.1: 44978379-44978379 on Build GRCh38
rs755654024 Chr.1: 44978345-44978345 on Build GRCh38
rs141988913 Chr.1: 44978337-44978337 on Build GRCh38
rs371483706 Chr.1: 44978482-44978482 on Build GRCh38
rs1002021284 Chr.1: 44978485-44978485 on Build GRCh38
rs754161506 Chr.1: 44978293-44978293 on Build GRCh38
rs754975429 Chr.1: 44978462-44978462 on Build GRCh38
rs144054571 Chr.1: 44978385-44978385 on Build GRCh38
rs549453394 Chr.1: 44978467-44978467 on Build GRCh38
rs374502902 Chr.1: 44978463-44978463 on Build GRCh38
rs747049484 Chr.1: 44978459-44978459 on Build GRCh38
rs757505662 Chr.1: 44978297-44978297 on Build GRCh38
rs778945531 Chr.1: 44978325-44978325 on Build GRCh38
rs772111952 Chr.1: 44978338-44978338 on Build GRCh38
rs757621916 Chr.1: 44978419-44978419 on Build GRCh38
rs371181750 Chr.1: 44978393-44978393 on Build GRCh38
rs761201445 Chr.1: 44978403-44978403 on Build GRCh38
rs72887005 Chr.1: 44978464-44978464 on Build GRCh38
rs113994022 Chr.1: 44978349-44978349 on Build GRCh38
rs776568212 Chr.1: 44978365-44978365 on Build GRCh38
rs770531078 Chr.1: 44978483-44978483 on Build GRCh38
rs768670174 Chr.1: 44978359-44978359 on Build GRCh38
rs910289205 Chr.1: 44978304-44978304 on Build GRCh38
rs773036534 Chr.1: 44978474-44978474 on Build GRCh38
rs758627166 Chr.1: 44978425-44978425 on Build GRCh38
rs774721437 Chr.1: 44978383-44978383 on Build GRCh38
rs765921585 Chr.1: 44978392-44978392 on Build GRCh38
rs754243775 Chr.1: 44978405-44978405 on Build GRCh38
rs142753066 Chr.1: 44978420-44978420 on Build GRCh38
rs201906174 Chr.1: 44978486-44978486 on Build GRCh38
rs764519183 Chr.1: 44978404-44978404 on Build GRCh38
rs780307542 Chr.1: 44978427-44978427 on Build GRCh38

View Full Product Details