Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236482204-236482473 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDARADD
Entrez Gene ID
128178
Gene Name
EDAR associated death domain
Gene Aliases
ECTD11A, ECTD11B, ED3, EDA3
UniGene
Hs.352224
Gene Chromosome Location
Chr.1: 236394380-236484708 on Build GRCh38
Transcripts
NM_080738.3 NM_145861.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
116919 c.382A>C p.K128Q Chr.1: 236482383-236482383 on Build GRCh38
1185796 c.404C>A p.T135K Chr.1: 236482405-236482405 on Build GRCh38
6062123 c.387G>T p.L129L Chr.1: 236482388-236482388 on Build GRCh38
6062125 c.328C>A p.P110T Chr.1: 236482329-236482329 on Build GRCh38
5827778 c.339C>T p.D113D Chr.1: 236482370-236482370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397515575 Chr.1: 236482403-236482408 on Build GRCh38
rs780289873 Chr.1: 236482253-236482253 on Build GRCh38
rs373971442 Chr.1: 236482283-236482283 on Build GRCh38
rs114632254 Chr.1: 236482309-236482309 on Build GRCh38
rs936740826 Chr.1: 236482338-236482338 on Build GRCh38
rs752275345 Chr.1: 236482324-236482324 on Build GRCh38
rs604070 Chr.1: 236482370-236482370 on Build GRCh38
rs769122607 Chr.1: 236482405-236482405 on Build GRCh38
rs760843174 Chr.1: 236482400-236482400 on Build GRCh38
rs369166930 Chr.1: 236482245-236482245 on Build GRCh38
rs758607876 Chr.1: 236482399-236482403 on Build GRCh38
rs376972393 Chr.1: 236482379-236482379 on Build GRCh38
rs147501905 Chr.1: 236482371-236482371 on Build GRCh38
rs954823206 Chr.1: 236482418-236482418 on Build GRCh38
rs761026610 Chr.1: 236482259-236482259 on Build GRCh38
rs139996586 Chr.1: 236482394-236482394 on Build GRCh38
rs779277524 Chr.1: 236482376-236482376 on Build GRCh38
rs376675770 Chr.1: 236482239-236482239 on Build GRCh38
rs1005957166 Chr.1: 236482393-236482393 on Build GRCh38
rs777899133 Chr.1: 236482356-236482356 on Build GRCh38
rs147249573 Chr.1: 236482323-236482323 on Build GRCh38
rs754505568 Chr.1: 236482322-236482322 on Build GRCh38
rs534068767 Chr.1: 236482279-236482279 on Build GRCh38
rs755966826 Chr.1: 236482350-236482350 on Build GRCh38
rs1031441787 Chr.1: 236482450-236482450 on Build GRCh38
rs759679168 Chr.1: 236482244-236482244 on Build GRCh38
rs906737114 Chr.1: 236482349-236482349 on Build GRCh38
rs1030214435 Chr.1: 236482414-236482414 on Build GRCh38
rs762203601 Chr.1: 236482295-236482295 on Build GRCh38
rs886046186 Chr.1: 236482303-236482303 on Build GRCh38
rs764680952 Chr.1: 236482261-236482261 on Build GRCh38
rs199965336 Chr.1: 236482430-236482430 on Build GRCh38
rs969366797 Chr.1: 236482311-236482311 on Build GRCh38
rs765992938 Chr.1: 236482299-236482299 on Build GRCh38
rs797045087 Chr.1: 236482300-236482301 on Build GRCh38
rs750512955 Chr.1: 236482267-236482268 on Build GRCh38
rs772349212 Chr.1: 236482392-236482392 on Build GRCh38
rs773579559 Chr.1: 236482454-236482454 on Build GRCh38
rs771250194 Chr.1: 236482233-236482233 on Build GRCh38
rs121908116 Chr.1: 236482366-236482366 on Build GRCh38
rs879255629 Chr.1: 236482368-236482368 on Build GRCh38
rs777172467 Chr.1: 236482406-236482406 on Build GRCh38
rs369614195 Chr.1: 236482451-236482451 on Build GRCh38

View Full Product Details