Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45136415-45136676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACBD4
Entrez Gene ID
79777
Gene Name
acyl-CoA binding domain containing 4
Gene Aliases
HMFT0700
UniGene
Hs.110298
Gene Chromosome Location
Chr.17: 45132600-45144181 on Build GRCh38
Transcripts
NM_001135705.2 NM_001135706.2 NM_001135707.2 NM_001321352.1 NM_001321353.1 NM_024722.3 XM_006722085.2 XM_011525259.2 XM_011525261.2 XM_017025084.1 XM_017025085.1 XM_017025086.1 XM_017025087.1 XM_017025088.1 XM_017025089.1 XM_017025090.1 XM_017025091.1 XM_017025092.1 XM_017025093.1 XM_017025094.1 XM_017025095.1 XM_017025096.1 XM_017025097.1 XM_017025098.1 XM_017025099.1 XM_017025100.1 XM_017025101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5816681 c.180G>T p.G60G Chr.17: 45136591-45136591 on Build GRCh38
5055529 c.205A>G p.K69E Chr.17: 45136616-45136616 on Build GRCh38
6556416 c.145G>T p.A49S Chr.17: 45136556-45136556 on Build GRCh38
79124 c.199C>T p.R67* Chr.17: 45136610-45136610 on Build GRCh38
980109 c.97C>T p.R33C Chr.17: 45136508-45136508 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757668347 Chr.17: 45136523-45136523 on Build GRCh38
rs750628420 Chr.17: 45136526-45136526 on Build GRCh38
rs765863543 Chr.17: 45136508-45136508 on Build GRCh38
rs748121568 Chr.17: 45136583-45136583 on Build GRCh38
rs569865532 Chr.17: 45136605-45136605 on Build GRCh38
rs377076251 Chr.17: 45136615-45136615 on Build GRCh38
rs377441547 Chr.17: 45136651-45136651 on Build GRCh38
rs754872454 Chr.17: 45136579-45136579 on Build GRCh38
rs771812922 Chr.17: 45136584-45136584 on Build GRCh38
rs369056152 Chr.17: 45136602-45136602 on Build GRCh38
rs756565451 Chr.17: 45136635-45136635 on Build GRCh38
rs201576973 Chr.17: 45136546-45136546 on Build GRCh38
rs769479084 Chr.17: 45136506-45136506 on Build GRCh38
rs771900960 Chr.17: 45136481-45136481 on Build GRCh38
rs767939172 Chr.17: 45136634-45136634 on Build GRCh38
rs774961769 Chr.17: 45136624-45136624 on Build GRCh38
rs760351023 Chr.17: 45136494-45136494 on Build GRCh38
rs781716222 Chr.17: 45136524-45136524 on Build GRCh38
rs577647382 Chr.17: 45136511-45136511 on Build GRCh38
rs774411250 Chr.17: 45136464-45136464 on Build GRCh38
rs375914906 Chr.17: 45136527-45136527 on Build GRCh38
rs759153871 Chr.17: 45136509-45136509 on Build GRCh38
rs761147266 Chr.17: 45136463-45136463 on Build GRCh38
rs937121767 Chr.17: 45136435-45136435 on Build GRCh38
rs773750392 Chr.17: 45136437-45136437 on Build GRCh38
rs200711802 Chr.17: 45136445-45136445 on Build GRCh38
rs935961977 Chr.17: 45136547-45136547 on Build GRCh38
rs370269279 Chr.17: 45136658-45136658 on Build GRCh38
rs944810405 Chr.17: 45136472-45136472 on Build GRCh38
rs776491369 Chr.17: 45136453-45136453 on Build GRCh38
rs373383277 Chr.17: 45136626-45136626 on Build GRCh38
rs766566117 Chr.17: 45136636-45136636 on Build GRCh38
rs768835046 Chr.17: 45136551-45136551 on Build GRCh38
rs770486647 Chr.17: 45136591-45136591 on Build GRCh38
rs780110670 Chr.17: 45136531-45136531 on Build GRCh38
rs764845745 Chr.17: 45136609-45136609 on Build GRCh38
rs796925582 Chr.17: 45136439-45136439 on Build GRCh38
rs749188178 Chr.17: 45136460-45136460 on Build GRCh38
rs772819405 Chr.17: 45136490-45136490 on Build GRCh38
rs367783132 Chr.17: 45136533-45136533 on Build GRCh38
rs761579747 Chr.17: 45136476-45136476 on Build GRCh38
rs761610565 Chr.17: 45136478-45136478 on Build GRCh38
rs760263177 Chr.17: 45136589-45136589 on Build GRCh38
rs752163773 Chr.17: 45136518-45136518 on Build GRCh38
rs778691797 Chr.17: 45136555-45136555 on Build GRCh38
rs370172638 Chr.17: 45136618-45136618 on Build GRCh38
rs376859282 Chr.17: 45136588-45136588 on Build GRCh38

View Full Product Details