Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1419876-1420149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf91
Entrez Gene ID
283951
Gene Name
chromosome 16 open reading frame 91
Gene Aliases
CCSMST1, URLC5, gs103
UniGene
Hs.58373
Gene Chromosome Location
Chr.16: 1419744-1420800 on Build GRCh38
Transcripts
NM_001272051.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7165831 c.921G>A p.V307V Chr.16: 1420093-1420093 on Build GRCh38
4836311 c.939C>T p.L313L Chr.16: 1420075-1420075 on Build GRCh38
967243 c.1019G>A p.R340K Chr.16: 1419995-1419995 on Build GRCh38
5012952 c.913A>C p.I305L Chr.16: 1420101-1420101 on Build GRCh38
6614625 c.930G>A p.E310E Chr.16: 1420084-1420084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375027680 Chr.16: 1419959-1419959 on Build GRCh38
rs753546665 Chr.16: 1420122-1420122 on Build GRCh38
rs771152430 Chr.16: 1419982-1419982 on Build GRCh38
rs748822788 Chr.16: 1420036-1420036 on Build GRCh38
rs769758000 Chr.16: 1420070-1420070 on Build GRCh38
rs755559978 Chr.16: 1420031-1420031 on Build GRCh38
rs554249720 Chr.16: 1419921-1419921 on Build GRCh38
rs748597268 Chr.16: 1419960-1419960 on Build GRCh38
rs775287516 Chr.16: 1420016-1420016 on Build GRCh38
rs767048866 Chr.16: 1420027-1420027 on Build GRCh38
rs139891753 Chr.16: 1420114-1420114 on Build GRCh38
rs758800893 Chr.16: 1420043-1420043 on Build GRCh38
rs760892985 Chr.16: 1420000-1420000 on Build GRCh38
rs202013438 Chr.16: 1420095-1420095 on Build GRCh38
rs755755353 Chr.16: 1419951-1419951 on Build GRCh38
rs775560459 Chr.16: 1420008-1420008 on Build GRCh38
rs771605741 Chr.16: 1420055-1420055 on Build GRCh38
rs780623913 Chr.16: 1419926-1419926 on Build GRCh38
rs765543314 Chr.16: 1420001-1420001 on Build GRCh38
rs747546128 Chr.16: 1420050-1420050 on Build GRCh38
rs918700600 Chr.16: 1419974-1419974 on Build GRCh38
rs751512211 Chr.16: 1420022-1420022 on Build GRCh38
rs909330261 Chr.16: 1419983-1419983 on Build GRCh38
rs745542641 Chr.16: 1419941-1419941 on Build GRCh38
rs779817250 Chr.16: 1420035-1420035 on Build GRCh38
rs776808972 Chr.16: 1419994-1419994 on Build GRCh38
rs767494470 Chr.16: 1420104-1420104 on Build GRCh38
rs879252309 Chr.16: 1420117-1420117 on Build GRCh38
rs755720419 Chr.16: 1420111-1420111 on Build GRCh38
rs775786979 Chr.16: 1420072-1420072 on Build GRCh38
rs749979500 Chr.16: 1420030-1420030 on Build GRCh38
rs762697035 Chr.16: 1420046-1420046 on Build GRCh38
rs756766613 Chr.16: 1419923-1419923 on Build GRCh38
rs372002222 Chr.16: 1419918-1419918 on Build GRCh38
rs773771066 Chr.16: 1419970-1419970 on Build GRCh38
rs771085138 Chr.16: 1420002-1420002 on Build GRCh38
rs904869320 Chr.16: 1420075-1420075 on Build GRCh38
rs747458223 Chr.16: 1419981-1419981 on Build GRCh38
rs759656820 Chr.16: 1419998-1419998 on Build GRCh38
rs889807423 Chr.16: 1420006-1420006 on Build GRCh38
rs763030219 Chr.16: 1420089-1420089 on Build GRCh38
rs757138439 Chr.16: 1420026-1420026 on Build GRCh38
rs777822936 Chr.16: 1419920-1419920 on Build GRCh38
rs762874024 Chr.16: 1420013-1420013 on Build GRCh38
rs35518497 Chr.16: 1420007-1420008 on Build GRCh38
rs758104379 Chr.16: 1419919-1419919 on Build GRCh38
rs114154456 Chr.16: 1420067-1420067 on Build GRCh38
rs1010857728 Chr.16: 1420077-1420077 on Build GRCh38
rs576836801 Chr.16: 1420098-1420098 on Build GRCh38
rs1007116505 Chr.16: 1420015-1420015 on Build GRCh38
rs764971797 Chr.16: 1419905-1419905 on Build GRCh38
rs146420700 Chr.16: 1420066-1420066 on Build GRCh38

View Full Product Details