Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2181381-2181638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASKIN1
Entrez Gene ID
57524
Gene Name
CASK interacting protein 1
Gene Aliases
ANKS5A
UniGene
Hs.643537
Gene Chromosome Location
Chr.16: 2177183-2196464 on Build GRCh38
Transcripts
NM_020764.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620510 c.1915C>T p.P639S Chr.16: 2181453-2181453 on Build GRCh38
2152796 c.1394C>T p.P465L Chr.16: 2181461-2181461 on Build GRCh38
3278649 c.1409C>T p.P470L Chr.16: 2181446-2181446 on Build GRCh38
4411610 c.1952C>T p.T651I Chr.16: 2181416-2181416 on Build GRCh38
4411611 c.1439C>T p.T480I Chr.16: 2181416-2181416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755765518 Chr.16: 2181538-2181538 on Build GRCh38
rs763106393 Chr.16: 2181617-2181617 on Build GRCh38
rs934188352 Chr.16: 2181520-2181520 on Build GRCh38
rs370499776 Chr.16: 2181461-2181461 on Build GRCh38
rs954391848 Chr.16: 2181517-2181517 on Build GRCh38
rs772374918 Chr.16: 2181464-2181464 on Build GRCh38
rs367596621 Chr.16: 2181486-2181486 on Build GRCh38
rs770027178 Chr.16: 2181475-2181477 on Build GRCh38
rs202188634 Chr.16: 2181459-2181459 on Build GRCh38
rs746699704 Chr.16: 2181447-2181447 on Build GRCh38
rs753397435 Chr.16: 2181556-2181556 on Build GRCh38
rs764096334 Chr.16: 2181526-2181526 on Build GRCh38
rs750636833 Chr.16: 2181416-2181416 on Build GRCh38
rs779483964 Chr.16: 2181545-2181545 on Build GRCh38
rs762091531 Chr.16: 2181403-2181403 on Build GRCh38
rs745480941 Chr.16: 2181499-2181499 on Build GRCh38
rs994094112 Chr.16: 2181610-2181610 on Build GRCh38
rs747375054 Chr.16: 2181512-2181512 on Build GRCh38
rs757061472 Chr.16: 2181493-2181493 on Build GRCh38
rs1030069400 Chr.16: 2181516-2181516 on Build GRCh38
rs374863564 Chr.16: 2181467-2181467 on Build GRCh38
rs528048966 Chr.16: 2181616-2181616 on Build GRCh38
rs772614722 Chr.16: 2181509-2181509 on Build GRCh38
rs751387277 Chr.16: 2181489-2181489 on Build GRCh38
rs374123215 Chr.16: 2181445-2181445 on Build GRCh38
rs760868396 Chr.16: 2181422-2181422 on Build GRCh38
rs577001878 Chr.16: 2181528-2181528 on Build GRCh38
rs753898630 Chr.16: 2181439-2181439 on Build GRCh38
rs762864482 Chr.16: 2181523-2181523 on Build GRCh38
rs989675498 Chr.16: 2181603-2181603 on Build GRCh38
rs1038352691 Chr.16: 2181431-2181431 on Build GRCh38
rs766509114 Chr.16: 2181430-2181430 on Build GRCh38
rs776764949 Chr.16: 2181474-2181474 on Build GRCh38
rs553437052 Chr.16: 2181466-2181466 on Build GRCh38
rs376676749 Chr.16: 2181513-2181513 on Build GRCh38
rs570994763 Chr.16: 2181460-2181460 on Build GRCh38
rs767886341 Chr.16: 2181415-2181415 on Build GRCh38
rs201714805 Chr.16: 2181481-2181481 on Build GRCh38
rs752639045 Chr.16: 2181442-2181442 on Build GRCh38
rs756965867 Chr.16: 2181454-2181454 on Build GRCh38
rs184044233 Chr.16: 2181503-2181503 on Build GRCh38
rs1029974663 Chr.16: 2181398-2181398 on Build GRCh38
rs1055672159 Chr.16: 2181498-2181498 on Build GRCh38
rs759641819 Chr.16: 2181475-2181475 on Build GRCh38
rs780908631 Chr.16: 2181456-2181456 on Build GRCh38
rs559388298 Chr.16: 2181611-2181611 on Build GRCh38
rs746135545 Chr.16: 2181609-2181609 on Build GRCh38
rs771326515 Chr.16: 2181605-2181605 on Build GRCh38
rs768567680 Chr.16: 2181619-2181619 on Build GRCh38
rs747477325 Chr.16: 2181592-2181592 on Build GRCh38
rs1037906253 Chr.16: 2181507-2181507 on Build GRCh38
rs368506468 Chr.16: 2181537-2181537 on Build GRCh38
rs758325357 Chr.16: 2181484-2181484 on Build GRCh38
rs200132110 Chr.16: 2181441-2181441 on Build GRCh38
rs776859432 Chr.16: 2181518-2181518 on Build GRCh38
rs986406188 Chr.16: 2181438-2181438 on Build GRCh38
rs781514977 Chr.16: 2181608-2181608 on Build GRCh38
rs780998571 Chr.16: 2181494-2181494 on Build GRCh38
rs757138859 Chr.16: 2181536-2181536 on Build GRCh38
rs538890179 Chr.16: 2181490-2181490 on Build GRCh38
rs544406031 Chr.16: 2181510-2181510 on Build GRCh38
rs778278356 Chr.16: 2181577-2181577 on Build GRCh38
rs556281885 Chr.16: 2181519-2181519 on Build GRCh38
rs572855009 Chr.16: 2181478-2181478 on Build GRCh38
rs555407612 Chr.16: 2181501-2181501 on Build GRCh38
rs773809082 Chr.16: 2181614-2181614 on Build GRCh38
rs201669676 Chr.16: 2181446-2181446 on Build GRCh38
rs779597891 Chr.16: 2181502-2181502 on Build GRCh38
rs766598978 Chr.16: 2181470-2181470 on Build GRCh38
rs754391474 Chr.16: 2181576-2181576 on Build GRCh38
rs752624487 Chr.16: 2181479-2181479 on Build GRCh38

View Full Product Details