Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 8735629-8735902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABAT
Entrez Gene ID
18
Gene Name
4-aminobutyrate aminotransferase
Gene Aliases
GABA-AT, GABAT, NPD009
UniGene
Hs.336768
Gene Chromosome Location
Chr.16: 8674587-8784575 on Build GRCh38
Transcripts
NM_000663.4 NM_001127448.1 NM_020686.5 XM_011522400.2 XM_011522401.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3513170 c.9C>T p.S3S Chr.16: 8735748-8735748 on Build GRCh38
435896 c.10A>G p.M4V Chr.16: 8735749-8735749 on Build GRCh38
704466 c.70+1G>T p.? Chr.16: 8735810-8735810 on Build GRCh38
7252741 c.55C>T p.R19C Chr.16: 8735794-8735794 on Build GRCh38
6589372 c.33C>T p.A11A Chr.16: 8735772-8735772 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs769650439 Chr.16: 8735724-8735724 on Build GRCh38
rs770877180 Chr.16: 8735670-8735670 on Build GRCh38
rs979943233 Chr.16: 8735652-8735652 on Build GRCh38
rs773255451 Chr.16: 8735759-8735759 on Build GRCh38
rs988166003 Chr.16: 8735876-8735876 on Build GRCh38
rs145493136 Chr.16: 8735869-8735869 on Build GRCh38
rs745997771 Chr.16: 8735709-8735709 on Build GRCh38
rs1021270177 Chr.16: 8735828-8735828 on Build GRCh38
rs371707643 Chr.16: 8735701-8735701 on Build GRCh38
rs375337638 Chr.16: 8735783-8735783 on Build GRCh38
rs754221172 Chr.16: 8735836-8735836 on Build GRCh38
rs757766788 Chr.16: 8735839-8735839 on Build GRCh38
rs779624302 Chr.16: 8735785-8735785 on Build GRCh38
rs77696190 Chr.16: 8735794-8735794 on Build GRCh38
rs553745013 Chr.16: 8735819-8735819 on Build GRCh38
rs531783337 Chr.16: 8735765-8735765 on Build GRCh38
rs763704300 Chr.16: 8735829-8735829 on Build GRCh38
rs927076888 Chr.16: 8735657-8735657 on Build GRCh38
rs987724126 Chr.16: 8735745-8735745 on Build GRCh38
rs34813662 Chr.16: 8735769-8735769 on Build GRCh38
rs770850099 Chr.16: 8735805-8735805 on Build GRCh38
rs773032873 Chr.16: 8735802-8735802 on Build GRCh38
rs369944447 Chr.16: 8735815-8735815 on Build GRCh38
rs778232601 Chr.16: 8735772-8735772 on Build GRCh38
rs536656603 Chr.16: 8735826-8735826 on Build GRCh38
rs766349160 Chr.16: 8735757-8735757 on Build GRCh38
rs767118285 Chr.16: 8735875-8735875 on Build GRCh38
rs141213694 Chr.16: 8735758-8735758 on Build GRCh38
rs762710511 Chr.16: 8735803-8735803 on Build GRCh38
rs765776223 Chr.16: 8735843-8735843 on Build GRCh38
rs201701679 Chr.16: 8735764-8735764 on Build GRCh38
rs539939852 Chr.16: 8735862-8735862 on Build GRCh38
rs867072039 Chr.16: 8735748-8735748 on Build GRCh38
rs768798452 Chr.16: 8735799-8735799 on Build GRCh38
rs762912862 Chr.16: 8735747-8735747 on Build GRCh38
rs374147109 Chr.16: 8735818-8735818 on Build GRCh38
rs551229015 Chr.16: 8735687-8735687 on Build GRCh38
rs552341323 Chr.16: 8735793-8735793 on Build GRCh38
rs138270964 Chr.16: 8735784-8735784 on Build GRCh38
rs904280183 Chr.16: 8735851-8735851 on Build GRCh38
rs780368959 Chr.16: 8735795-8735795 on Build GRCh38
rs758509143 Chr.16: 8735854-8735854 on Build GRCh38
rs1640998 Chr.16: 8735858-8735858 on Build GRCh38
rs866966999 Chr.16: 8735827-8735827 on Build GRCh38
rs200472536 Chr.16: 8735710-8735710 on Build GRCh38
rs532952970 Chr.16: 8735737-8735737 on Build GRCh38
rs747283357 Chr.16: 8735797-8735797 on Build GRCh38
rs369376388 Chr.16: 8735807-8735807 on Build GRCh38
rs761451655 Chr.16: 8735713-8735713 on Build GRCh38
rs960270293 Chr.16: 8735680-8735680 on Build GRCh38
rs750983247 Chr.16: 8735846-8735846 on Build GRCh38
rs760345321 Chr.16: 8735821-8735821 on Build GRCh38
rs867427761 Chr.16: 8735703-8735703 on Build GRCh38
rs955140198 Chr.16: 8735711-8735711 on Build GRCh38
rs535494372 Chr.16: 8735817-8735817 on Build GRCh38
rs780248403 Chr.16: 8735856-8735856 on Build GRCh38
rs191784275 Chr.16: 8735873-8735873 on Build GRCh38
rs774967432 Chr.16: 8735820-8735820 on Build GRCh38

View Full Product Details