Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 29071716-29071989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPVL
Entrez Gene ID
54504
Gene Name
carboxypeptidase, vitellogenic like
Gene Aliases
HVLP
UniGene
Hs.233389
Gene Chromosome Location
Chr.7: 28995630-29146537 on Build GRCh38
Transcripts
NM_019029.2 NM_031311.3 XM_011515437.1 XM_017012366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
69329 c.864+2T>G p.? Chr.7: 29071771-29071771 on Build GRCh38
1725416 c.793A>G p.K265E Chr.7: 29071844-29071844 on Build GRCh38
5014677 c.778G>A p.D260N Chr.7: 29071859-29071859 on Build GRCh38
6442623 c.848G>A p.W283* Chr.7: 29071789-29071789 on Build GRCh38
3027699 c.794delA p.K265fs*11 Chr.7: 29071843-29071843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141896273 Chr.7: 29071808-29071808 on Build GRCh38
rs751839329 Chr.7: 29071878-29071878 on Build GRCh38
rs913045614 Chr.7: 29071893-29071894 on Build GRCh38
rs146943108 Chr.7: 29071909-29071909 on Build GRCh38
rs773783676 Chr.7: 29071946-29071946 on Build GRCh38
rs988704519 Chr.7: 29071840-29071840 on Build GRCh38
rs779903610 Chr.7: 29071927-29071927 on Build GRCh38
rs369462489 Chr.7: 29071817-29071817 on Build GRCh38
rs781658305 Chr.7: 29071959-29071959 on Build GRCh38
rs994625665 Chr.7: 29071807-29071807 on Build GRCh38
rs763570946 Chr.7: 29071769-29071769 on Build GRCh38
rs760391177 Chr.7: 29071763-29071763 on Build GRCh38
rs2074784 Chr.7: 29071962-29071962 on Build GRCh38
rs745827507 Chr.7: 29071755-29071755 on Build GRCh38
rs138534158 Chr.7: 29071925-29071925 on Build GRCh38
rs56680259 Chr.7: 29071930-29071930 on Build GRCh38
rs376702592 Chr.7: 29071828-29071828 on Build GRCh38
rs753228022 Chr.7: 29071830-29071830 on Build GRCh38
rs766988574 Chr.7: 29071921-29071931 on Build GRCh38
rs776498400 Chr.7: 29071754-29071754 on Build GRCh38
rs761413761 Chr.7: 29071775-29071775 on Build GRCh38
rs770884548 Chr.7: 29071749-29071749 on Build GRCh38
rs755368190 Chr.7: 29071827-29071827 on Build GRCh38
rs769675787 Chr.7: 29071757-29071757 on Build GRCh38
rs761299787 Chr.7: 29071950-29071950 on Build GRCh38
rs201462882 Chr.7: 29071834-29071834 on Build GRCh38
rs200288084 Chr.7: 29071833-29071833 on Build GRCh38
rs540733056 Chr.7: 29071895-29071895 on Build GRCh38
rs775490579 Chr.7: 29071758-29071758 on Build GRCh38
rs768185823 Chr.7: 29071940-29071940 on Build GRCh38
rs201079331 Chr.7: 29071772-29071772 on Build GRCh38
rs766744353 Chr.7: 29071796-29071796 on Build GRCh38
rs900622823 Chr.7: 29071924-29071924 on Build GRCh38
rs745714553 Chr.7: 29071916-29071916 on Build GRCh38

View Full Product Details