Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27558502-27558749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf72
Entrez Gene ID
203228
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
UniGene
Hs.493639
Gene Chromosome Location
Chr.9: 27546546-27573866 on Build GRCh38
Transcripts
NM_001256054.2 NM_018325.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6517025 c.805G>T p.E269* Chr.9: 27558541-27558541 on Build GRCh38
380700 c.796T>G p.C266G Chr.9: 27558550-27558550 on Build GRCh38
77404 c.766A>G p.T256A Chr.9: 27558580-27558580 on Build GRCh38
1461904 c.743T>C p.V248A Chr.9: 27558603-27558603 on Build GRCh38
6473117 c.822T>C p.Y274Y Chr.9: 27558524-27558524 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749267596 Chr.9: 27558648-27558648 on Build GRCh38
rs778084380 Chr.9: 27558594-27558594 on Build GRCh38
rs781544831 Chr.9: 27558611-27558611 on Build GRCh38
rs563678519 Chr.9: 27558660-27558660 on Build GRCh38
rs886063845 Chr.9: 27558554-27558554 on Build GRCh38
rs747748716 Chr.9: 27558602-27558602 on Build GRCh38
rs745871798 Chr.9: 27558616-27558616 on Build GRCh38
rs965935262 Chr.9: 27558615-27558615 on Build GRCh38
rs775518223 Chr.9: 27558636-27558636 on Build GRCh38
rs765663452 Chr.9: 27558564-27558564 on Build GRCh38
rs749883850 Chr.9: 27558547-27558547 on Build GRCh38
rs771670543 Chr.9: 27558666-27558666 on Build GRCh38
rs549517242 Chr.9: 27558681-27558681 on Build GRCh38
rs1052136982 Chr.9: 27558589-27558589 on Build GRCh38
rs758724401 Chr.9: 27558581-27558581 on Build GRCh38
rs753228403 Chr.9: 27558569-27558569 on Build GRCh38
rs530889918 Chr.9: 27558675-27558675 on Build GRCh38
rs747458864 Chr.9: 27558605-27558605 on Build GRCh38
rs757408861 Chr.9: 27558607-27558607 on Build GRCh38
rs755714382 Chr.9: 27558550-27558550 on Build GRCh38
rs767245865 Chr.9: 27558544-27558544 on Build GRCh38
rs770008724 Chr.9: 27558623-27558623 on Build GRCh38
rs774015166 Chr.9: 27558533-27558533 on Build GRCh38
rs768754064 Chr.9: 27558652-27558652 on Build GRCh38
rs749858364 Chr.9: 27558562-27558562 on Build GRCh38
rs545433336 Chr.9: 27558537-27558537 on Build GRCh38

View Full Product Details