Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154435067-154435332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6AP1
Entrez Gene ID
537
Gene Name
ATPase H+ transporting accessory protein 1
Gene Aliases
16A, ATP6IP1, ATP6S1, Ac45, CF2, VATPS1, XAP-3, XAP3
UniGene
Hs.633817
Gene Chromosome Location
Chr.X: 154428632-154436517 on Build GRCh38
Transcripts
NM_001183.5 XM_011531179.1

SNP Detail

SNP ID Chromosome Location
rs891593072 Chr.X: 154435159-154435159 on Build GRCh38
rs782248279 Chr.X: 154435267-154435267 on Build GRCh38
rs73638280 Chr.X: 154435112-154435112 on Build GRCh38
rs200790223 Chr.X: 154435111-154435111 on Build GRCh38
rs782144918 Chr.X: 154435123-154435123 on Build GRCh38
rs782323227 Chr.X: 154435094-154435094 on Build GRCh38
rs1012080406 Chr.X: 154435235-154435235 on Build GRCh38
rs781834817 Chr.X: 154435199-154435199 on Build GRCh38
rs782744522 Chr.X: 154435197-154435197 on Build GRCh38
rs782243961 Chr.X: 154435293-154435293 on Build GRCh38
rs371934768 Chr.X: 154435260-154435260 on Build GRCh38
rs143004832 Chr.X: 154435313-154435313 on Build GRCh38
rs138927310 Chr.X: 154435142-154435142 on Build GRCh38
rs781847623 Chr.X: 154435145-154435145 on Build GRCh38
rs782330877 Chr.X: 154435291-154435291 on Build GRCh38
rs782375587 Chr.X: 154435103-154435103 on Build GRCh38
rs782344183 Chr.X: 154435097-154435097 on Build GRCh38
rs782087266 Chr.X: 154435116-154435116 on Build GRCh38
rs878853278 Chr.X: 154435153-154435153 on Build GRCh38
rs1008076942 Chr.X: 154435121-154435121 on Build GRCh38
rs782447719 Chr.X: 154435207-154435207 on Build GRCh38
rs367899064 Chr.X: 154435117-154435117 on Build GRCh38
rs782264629 Chr.X: 154435256-154435256 on Build GRCh38
rs782186939 Chr.X: 154435227-154435228 on Build GRCh38
rs782180502 Chr.X: 154435277-154435277 on Build GRCh38
rs782554726 Chr.X: 154435237-154435237 on Build GRCh38
rs782126375 Chr.X: 154435148-154435148 on Build GRCh38
rs375162515 Chr.X: 154435290-154435290 on Build GRCh38
rs781923057 Chr.X: 154435122-154435122 on Build GRCh38
rs782384749 Chr.X: 154435312-154435312 on Build GRCh38

View Full Product Details