Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99365798-99366041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARPC1A
Entrez Gene ID
10552
Gene Name
actin related protein 2/3 complex subunit 1A
Gene Aliases
Arc40, HEL-68, HEL-S-307, SOP2Hs, SOP2L
UniGene
Hs.124126
Gene Chromosome Location
Chr.7: 99325873-99366262 on Build GRCh38
Transcripts
NM_001190996.1 NM_006409.3

SNP Detail

SNP ID Chromosome Location
rs763793970 Chr.7: 99365845-99365845 on Build GRCh38
rs761340148 Chr.7: 99365866-99365866 on Build GRCh38
rs998171963 Chr.7: 99365937-99365937 on Build GRCh38
rs949080163 Chr.7: 99365994-99365994 on Build GRCh38
rs766972025 Chr.7: 99365996-99365996 on Build GRCh38
rs745977698 Chr.7: 99365951-99365951 on Build GRCh38
rs374494395 Chr.7: 99365971-99365971 on Build GRCh38
rs377614883 Chr.7: 99365932-99365932 on Build GRCh38
rs953145790 Chr.7: 99365890-99365890 on Build GRCh38
rs188648717 Chr.7: 99365918-99365918 on Build GRCh38
rs532195296 Chr.7: 99365987-99365987 on Build GRCh38
rs117765427 Chr.7: 99366000-99366000 on Build GRCh38
rs761546189 Chr.7: 99365976-99365976 on Build GRCh38
rs748408683 Chr.7: 99365958-99365958 on Build GRCh38
rs371311827 Chr.7: 99365965-99365965 on Build GRCh38
rs369737583 Chr.7: 99365846-99365846 on Build GRCh38
rs1044916586 Chr.7: 99365966-99365966 on Build GRCh38
rs916319809 Chr.7: 99365993-99365993 on Build GRCh38
rs772594251 Chr.7: 99365954-99365954 on Build GRCh38
rs374155785 Chr.7: 99365904-99365904 on Build GRCh38
rs755155389 Chr.7: 99365919-99365919 on Build GRCh38
rs984482950 Chr.7: 99365897-99365897 on Build GRCh38
rs113174237 Chr.7: 99366003-99366003 on Build GRCh38
rs758323414 Chr.7: 99365915-99365915 on Build GRCh38
rs376476555 Chr.7: 99365821-99365821 on Build GRCh38
rs990503040 Chr.7: 99365986-99365986 on Build GRCh38
rs771197650 Chr.7: 99365946-99365946 on Build GRCh38
rs779224360 Chr.7: 99365945-99365945 on Build GRCh38
rs549648767 Chr.7: 99365972-99365972 on Build GRCh38
rs764800793 Chr.7: 99365896-99365896 on Build GRCh38
rs571173529 Chr.7: 99365975-99365975 on Build GRCh38
rs777900143 Chr.7: 99365929-99365929 on Build GRCh38
rs754781207 Chr.7: 99365926-99365926 on Build GRCh38
rs775895253 Chr.7: 99365841-99365841 on Build GRCh38
rs965341869 Chr.7: 99365931-99365931 on Build GRCh38
rs761156018 Chr.7: 99365842-99365842 on Build GRCh38
rs982182598 Chr.7: 99365995-99365995 on Build GRCh38

View Full Product Details