Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48707446-48707681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYSLTR2
Entrez Gene ID
57105
Gene Name
cysteinyl leukotriene receptor 2
Gene Aliases
CYSLT2, CYSLT2R, GPCR21, HG57, HPN321, KPG_011, PSEC0146, hGPCR21
UniGene
Hs.253706
Gene Chromosome Location
Chr.13: 48653711-48709362 on Build GRCh38
Transcripts
NM_001308465.1 NM_001308467.1 NM_001308468.1 NM_001308469.1 NM_001308470.1 NM_001308471.1 NM_001308476.1 NM_020377.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4504421 c.664C>T p.L222L Chr.13: 48707481-48707481 on Build GRCh38
5842611 c.640delT p.F215fs*9 Chr.13: 48707457-48707457 on Build GRCh38
1367346 c.816G>A p.T272T Chr.13: 48707633-48707633 on Build GRCh38
1367345 c.707C>T p.S236L Chr.13: 48707524-48707524 on Build GRCh38
696466 c.708G>T p.S236S Chr.13: 48707525-48707525 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780341015 Chr.13: 48707491-48707491 on Build GRCh38
rs953527715 Chr.13: 48707481-48707481 on Build GRCh38
rs747743936 Chr.13: 48707618-48707618 on Build GRCh38
rs200631066 Chr.13: 48707592-48707592 on Build GRCh38
rs747690569 Chr.13: 48707497-48707497 on Build GRCh38
rs983349374 Chr.13: 48707546-48707546 on Build GRCh38
rs907773090 Chr.13: 48707550-48707550 on Build GRCh38
rs772464555 Chr.13: 48707636-48707640 on Build GRCh38
rs200866083 Chr.13: 48707568-48707568 on Build GRCh38
rs200349706 Chr.13: 48707631-48707631 on Build GRCh38
rs61735175 Chr.13: 48707524-48707524 on Build GRCh38
rs199523415 Chr.13: 48707612-48707612 on Build GRCh38
rs774050943 Chr.13: 48707581-48707583 on Build GRCh38
rs756485097 Chr.13: 48707490-48707490 on Build GRCh38
rs138229519 Chr.13: 48707579-48707579 on Build GRCh38
rs879177079 Chr.13: 48707526-48707526 on Build GRCh38
rs755327589 Chr.13: 48707609-48707609 on Build GRCh38
rs778434472 Chr.13: 48707615-48707615 on Build GRCh38
rs201860597 Chr.13: 48707573-48707573 on Build GRCh38
rs777488065 Chr.13: 48707622-48707622 on Build GRCh38
rs139465979 Chr.13: 48707562-48707562 on Build GRCh38
rs750866173 Chr.13: 48707486-48707486 on Build GRCh38
rs757281416 Chr.13: 48707471-48707471 on Build GRCh38
rs745499497 Chr.13: 48707630-48707632 on Build GRCh38
rs776586024 Chr.13: 48707633-48707633 on Build GRCh38
rs200656296 Chr.13: 48707525-48707525 on Build GRCh38
rs150160388 Chr.13: 48707627-48707627 on Build GRCh38
rs935458798 Chr.13: 48707634-48707634 on Build GRCh38
rs199903030 Chr.13: 48707533-48707533 on Build GRCh38
rs199534318 Chr.13: 48707494-48707494 on Build GRCh38
rs758252119 Chr.13: 48707652-48707652 on Build GRCh38
rs140113993 Chr.13: 48707476-48707476 on Build GRCh38
rs201590235 Chr.13: 48707549-48707549 on Build GRCh38
rs201827922 Chr.13: 48707511-48707511 on Build GRCh38
rs61735169 Chr.13: 48707543-48707543 on Build GRCh38
rs201815757 Chr.13: 48707532-48707532 on Build GRCh38
rs750488514 Chr.13: 48707575-48707575 on Build GRCh38
rs758106094 Chr.13: 48707619-48707619 on Build GRCh38
rs754174807 Chr.13: 48707608-48707608 on Build GRCh38
rs148425155 Chr.13: 48707632-48707632 on Build GRCh38
rs906249010 Chr.13: 48707645-48707645 on Build GRCh38
rs199927379 Chr.13: 48707567-48707567 on Build GRCh38
rs778578042 Chr.13: 48707496-48707496 on Build GRCh38
rs138722135 Chr.13: 48707649-48707649 on Build GRCh38
rs200593429 Chr.13: 48707504-48707504 on Build GRCh38
rs149666471 Chr.13: 48707605-48707605 on Build GRCh38
rs373607915 Chr.13: 48707636-48707636 on Build GRCh38
rs749662416 Chr.13: 48707493-48707493 on Build GRCh38

View Full Product Details