Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65881360-65881609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSW
Entrez Gene ID
1521
Gene Name
cathepsin W
Gene Aliases
LYPN
UniGene
Hs.416848
Gene Chromosome Location
Chr.11: 65879813-65883741 on Build GRCh38
Transcripts
NM_001335.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1202647 c.185G>A p.R62H Chr.11: 65881419-65881419 on Build GRCh38
2166611 c.203A>C p.H68P Chr.11: 65881437-65881437 on Build GRCh38
5477758 c.277G>T p.D93Y Chr.11: 65881511-65881511 on Build GRCh38
6519254 c.246C>A p.G82G Chr.11: 65881480-65881480 on Build GRCh38
4611560 c.230_232delAGG p.E79delE Chr.11: 65881464-65881466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140363156 Chr.11: 65881450-65881450 on Build GRCh38
rs750571192 Chr.11: 65881403-65881403 on Build GRCh38
rs763974974 Chr.11: 65881464-65881466 on Build GRCh38
rs770809672 Chr.11: 65881428-65881428 on Build GRCh38
rs756330033 Chr.11: 65881530-65881530 on Build GRCh38
rs143555562 Chr.11: 65881452-65881452 on Build GRCh38
rs781750007 Chr.11: 65881395-65881395 on Build GRCh38
rs141204630 Chr.11: 65881424-65881424 on Build GRCh38
rs1046157329 Chr.11: 65881413-65881413 on Build GRCh38
rs755179255 Chr.11: 65881563-65881563 on Build GRCh38
rs7122964 Chr.11: 65881531-65881531 on Build GRCh38
rs762205028 Chr.11: 65881502-65881502 on Build GRCh38
rs948272375 Chr.11: 65881503-65881503 on Build GRCh38
rs763750031 Chr.11: 65881475-65881475 on Build GRCh38
rs145070075 Chr.11: 65881427-65881427 on Build GRCh38
rs544107680 Chr.11: 65881417-65881417 on Build GRCh38
rs200649581 Chr.11: 65881441-65881441 on Build GRCh38
rs773950358 Chr.11: 65881543-65881543 on Build GRCh38
rs150054992 Chr.11: 65881418-65881418 on Build GRCh38
rs559701819 Chr.11: 65881574-65881574 on Build GRCh38
rs768186594 Chr.11: 65881508-65881508 on Build GRCh38
rs776521640 Chr.11: 65881431-65881431 on Build GRCh38
rs774609398 Chr.11: 65881479-65881479 on Build GRCh38
rs756238303 Chr.11: 65881405-65881405 on Build GRCh38
rs750952674 Chr.11: 65881520-65881520 on Build GRCh38
rs1013715587 Chr.11: 65881385-65881385 on Build GRCh38
rs145203884 Chr.11: 65881419-65881419 on Build GRCh38
rs371072395 Chr.11: 65881386-65881386 on Build GRCh38
rs749460950 Chr.11: 65881437-65881437 on Build GRCh38

View Full Product Details