Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140547687-140547930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKHD1-EIF4EBP3
Entrez Gene ID
404734
Gene Name
ANKHD1-EIF4EBP3 readthrough
Gene Aliases
MASK-BP3, MASK-BP3ARF
UniGene
Hs.594084
Gene Chromosome Location
Chr.5: 140401814-140549578 on Build GRCh38
Transcripts
NM_020690.5 NM_003732.2

Gene Symbol
EIF4EBP3
Entrez Gene ID
8637
Gene Name
eukaryotic translation initiation factor 4E binding protein 3
Gene Aliases
4E-BP3, 4EBP3
UniGene
Hs.594084
Gene Chromosome Location
Chr.5: 140547666-140549578 on Build GRCh38
Transcripts
NM_020690.5 NM_003732.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4966577 c.43C>T p.Q15* Chr.5: 140547780-140547780 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753147013 Chr.5: 140547711-140547711 on Build GRCh38
rs971559251 Chr.5: 140547877-140547877 on Build GRCh38
rs971447493 Chr.5: 140547869-140547869 on Build GRCh38
rs909536381 Chr.5: 140547714-140547714 on Build GRCh38
rs779561737 Chr.5: 140547790-140547790 on Build GRCh38
rs1005886719 Chr.5: 140547767-140547767 on Build GRCh38
rs924864764 Chr.5: 140547889-140547889 on Build GRCh38
rs781551674 Chr.5: 140547772-140547772 on Build GRCh38
rs959388671 Chr.5: 140547800-140547800 on Build GRCh38
rs1008149949 Chr.5: 140547719-140547734 on Build GRCh38
rs1050906174 Chr.5: 140547750-140547750 on Build GRCh38
rs1047667714 Chr.5: 140547709-140547709 on Build GRCh38
rs75475787 Chr.5: 140547830-140547830 on Build GRCh38
rs748460833 Chr.5: 140547835-140547835 on Build GRCh38
rs918417034 Chr.5: 140547731-140547731 on Build GRCh38
rs749837015 Chr.5: 140547745-140547745 on Build GRCh38
rs186301886 Chr.5: 140547786-140547786 on Build GRCh38
rs1020335138 Chr.5: 140547867-140547867 on Build GRCh38
rs1054879786 Chr.5: 140547816-140547816 on Build GRCh38
rs116513628 Chr.5: 140547851-140547851 on Build GRCh38
rs568288661 Chr.5: 140547873-140547873 on Build GRCh38
rs1006800770 Chr.5: 140547798-140547798 on Build GRCh38
rs778357350 Chr.5: 140547743-140547743 on Build GRCh38
rs981684955 Chr.5: 140547887-140547887 on Build GRCh38
rs1034463114 Chr.5: 140547880-140547880 on Build GRCh38
rs544983486 Chr.5: 140547734-140547734 on Build GRCh38
rs886128648 Chr.5: 140547738-140547738 on Build GRCh38
rs1016052711 Chr.5: 140547791-140547791 on Build GRCh38
rs200051899 Chr.5: 140547817-140547817 on Build GRCh38
rs933777214 Chr.5: 140547746-140547746 on Build GRCh38
rs372216475 Chr.5: 140547774-140547774 on Build GRCh38
rs1036758296 Chr.5: 140547728-140547728 on Build GRCh38

View Full Product Details