Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 75060252-75060513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB7
Entrez Gene ID
22
Gene Name
ATP binding cassette subfamily B member 7
Gene Aliases
ABC7, ASAT, Atm1p, EST140535
UniGene
Hs.370480
Gene Chromosome Location
Chr.X: 75053172-75156340 on Build GRCh38
Transcripts
NM_001271696.1 NM_001271697.1 NM_001271698.1 NM_001271699.1 NM_004299.4

SNP Detail

SNP ID Chromosome Location
rs968953459 Chr.X: 75060427-75060428 on Build GRCh38
rs367870900 Chr.X: 75060316-75060316 on Build GRCh38
rs924784945 Chr.X: 75060482-75060482 on Build GRCh38
rs145497613 Chr.X: 75060406-75060406 on Build GRCh38
rs1015402539 Chr.X: 75060386-75060386 on Build GRCh38
rs778468842 Chr.X: 75060373-75060373 on Build GRCh38
rs745524396 Chr.X: 75060439-75060440 on Build GRCh38
rs750737191 Chr.X: 75060360-75060360 on Build GRCh38
rs751648765 Chr.X: 75060315-75060315 on Build GRCh38
rs923541584 Chr.X: 75060441-75060441 on Build GRCh38
rs952109184 Chr.X: 75060337-75060337 on Build GRCh38
rs371969849 Chr.X: 75060348-75060348 on Build GRCh38
rs181336743 Chr.X: 75060371-75060371 on Build GRCh38
rs758041116 Chr.X: 75060375-75060375 on Build GRCh38
rs960064116 Chr.X: 75060351-75060351 on Build GRCh38
rs1004290081 Chr.X: 75060289-75060289 on Build GRCh38
rs747829914 Chr.X: 75060374-75060374 on Build GRCh38
rs113544170 Chr.X: 75060448-75060449 on Build GRCh38
rs953609941 Chr.X: 75060465-75060465 on Build GRCh38
rs185203869 Chr.X: 75060390-75060390 on Build GRCh38

View Full Product Details