Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 142914825-142915052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11B2
Entrez Gene ID
1585
Gene Name
cytochrome P450 family 11 subfamily B member 2
Gene Aliases
ALDOS, CPN2, CYP11B, CYP11BL, CYPXIB2, P-450C18, P450C18, P450aldo
UniGene
Hs.632054
Gene Chromosome Location
Chr.8: 142910559-142917843 on Build GRCh38
Transcripts
NM_000498.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1096893 c.622C>T p.R208W Chr.8: 142914882-142914882 on Build GRCh38
6180252 c.654T>G p.S218S Chr.8: 142914850-142914850 on Build GRCh38
5467366 c.619G>T p.E207* Chr.8: 142914885-142914885 on Build GRCh38
2867838 c.623G>A p.R208Q Chr.8: 142914881-142914881 on Build GRCh38
4675862 c.634G>T p.V212F Chr.8: 142914870-142914870 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777459030 Chr.8: 142914882-142914882 on Build GRCh38
rs746405641 Chr.8: 142915010-142915010 on Build GRCh38
rs747656269 Chr.8: 142914918-142914918 on Build GRCh38
rs768822913 Chr.8: 142914926-142914926 on Build GRCh38
rs757709071 Chr.8: 142915001-142915001 on Build GRCh38
rs6436 Chr.8: 142915026-142915026 on Build GRCh38
rs375189544 Chr.8: 142915003-142915003 on Build GRCh38
rs551933154 Chr.8: 142914864-142914864 on Build GRCh38
rs775388813 Chr.8: 142914913-142914913 on Build GRCh38
rs749398513 Chr.8: 142915019-142915019 on Build GRCh38
rs746315718 Chr.8: 142914909-142914909 on Build GRCh38
rs370388708 Chr.8: 142914912-142914912 on Build GRCh38
rs781765416 Chr.8: 142914902-142914902 on Build GRCh38
rs1009106764 Chr.8: 142914985-142914985 on Build GRCh38
rs753710760 Chr.8: 142914851-142914851 on Build GRCh38
rs370796995 Chr.8: 142914951-142914951 on Build GRCh38
rs777354935 Chr.8: 142914995-142914995 on Build GRCh38
rs890579892 Chr.8: 142914967-142914967 on Build GRCh38
rs923890288 Chr.8: 142914911-142914911 on Build GRCh38
rs759431398 Chr.8: 142914950-142914950 on Build GRCh38
rs757912502 Chr.8: 142914953-142914953 on Build GRCh38
rs113284476 Chr.8: 142914898-142914898 on Build GRCh38
rs6414 Chr.8: 142915018-142915018 on Build GRCh38
rs774633465 Chr.8: 142914931-142914931 on Build GRCh38
rs751188918 Chr.8: 142914998-142914998 on Build GRCh38
rs886062743 Chr.8: 142915031-142915031 on Build GRCh38
rs766374032 Chr.8: 142914848-142914848 on Build GRCh38
rs6416 Chr.8: 142914984-142914984 on Build GRCh38
rs760469914 Chr.8: 142914946-142914946 on Build GRCh38
rs796771655 Chr.8: 142914947-142914949 on Build GRCh38
rs570692763 Chr.8: 142914901-142914901 on Build GRCh38
rs778346287 Chr.8: 142914869-142914869 on Build GRCh38
rs749800670 Chr.8: 142914881-142914881 on Build GRCh38
rs760453419 Chr.8: 142914844-142914844 on Build GRCh38
rs6399 Chr.8: 142914949-142914949 on Build GRCh38
rs368519861 Chr.8: 142915017-142915017 on Build GRCh38
rs752426508 Chr.8: 142914874-142914874 on Build GRCh38
rs6418 Chr.8: 142914947-142914947 on Build GRCh38
rs763053942 Chr.8: 142914921-142914921 on Build GRCh38
rs6415 Chr.8: 142914993-142914993 on Build GRCh38
rs5307 Chr.8: 142915032-142915032 on Build GRCh38
rs747390303 Chr.8: 142914883-142914883 on Build GRCh38
rs6413 Chr.8: 142915033-142915033 on Build GRCh38

View Full Product Details