Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44625484-44625743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADA
Entrez Gene ID
100
Gene Name
adenosine deaminase
Gene Aliases
-
UniGene
Hs.654536
Gene Chromosome Location
Chr.20: 44619519-44651758 on Build GRCh38
Transcripts
NM_000022.3 NM_001322050.1 NM_001322051.1 NR_136160.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6417197 c.404A>G p.Q135R Chr.20: 44625643-44625643 on Build GRCh38
1734543 c.421delG p.E141fs*38 Chr.20: 44625626-44625626 on Build GRCh38
84729 c.425G>A p.R142Q Chr.20: 44625622-44625622 on Build GRCh38
1411974 c.390G>A p.V130V Chr.20: 44625657-44625657 on Build GRCh38
5857952 c.473A>G p.Q158R Chr.20: 44625574-44625574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778434803 Chr.20: 44625522-44625522 on Build GRCh38
rs766539394 Chr.20: 44625672-44625672 on Build GRCh38
rs551821715 Chr.20: 44625613-44625613 on Build GRCh38
rs121908728 Chr.20: 44625593-44625593 on Build GRCh38
rs750462837 Chr.20: 44625562-44625562 on Build GRCh38
rs61737144 Chr.20: 44625657-44625657 on Build GRCh38
rs536103858 Chr.20: 44625673-44625673 on Build GRCh38
rs1005504755 Chr.20: 44625691-44625691 on Build GRCh38
rs367574074 Chr.20: 44625696-44625696 on Build GRCh38
rs775400163 Chr.20: 44625538-44625538 on Build GRCh38
rs746917604 Chr.20: 44625629-44625629 on Build GRCh38
rs1005866894 Chr.20: 44625510-44625510 on Build GRCh38
rs752159265 Chr.20: 44625559-44625564 on Build GRCh38
rs767782363 Chr.20: 44625683-44625683 on Build GRCh38
rs868364372 Chr.20: 44625620-44625620 on Build GRCh38
rs61732239 Chr.20: 44625622-44625622 on Build GRCh38
rs768474992 Chr.20: 44625542-44625542 on Build GRCh38
rs121908737 Chr.20: 44625601-44625601 on Build GRCh38
rs121908735 Chr.20: 44625581-44625581 on Build GRCh38
rs990121469 Chr.20: 44625595-44625595 on Build GRCh38
rs760520726 Chr.20: 44625539-44625539 on Build GRCh38
rs1015455929 Chr.20: 44625692-44625692 on Build GRCh38
rs572092463 Chr.20: 44625663-44625663 on Build GRCh38
rs776757485 Chr.20: 44625546-44625546 on Build GRCh38
rs146921882 Chr.20: 44625645-44625645 on Build GRCh38
rs762046214 Chr.20: 44625549-44625549 on Build GRCh38
rs758073965 Chr.20: 44625680-44625681 on Build GRCh38
rs374116181 Chr.20: 44625534-44625534 on Build GRCh38
rs369538109 Chr.20: 44625554-44625554 on Build GRCh38
rs761242509 Chr.20: 44625568-44625568 on Build GRCh38
rs371028908 Chr.20: 44625589-44625589 on Build GRCh38
rs752094197 Chr.20: 44625576-44625576 on Build GRCh38
rs780014431 Chr.20: 44625623-44625623 on Build GRCh38
rs756427277 Chr.20: 44625695-44625695 on Build GRCh38
rs7344760 Chr.20: 44625666-44625666 on Build GRCh38
rs753035656 Chr.20: 44625687-44625687 on Build GRCh38
rs773557479 Chr.20: 44625665-44625665 on Build GRCh38
rs767053462 Chr.20: 44625571-44625571 on Build GRCh38
rs374413123 Chr.20: 44625615-44625615 on Build GRCh38
rs121908731 Chr.20: 44625662-44625662 on Build GRCh38
rs121908732 Chr.20: 44625628-44625628 on Build GRCh38
rs377753578 Chr.20: 44625535-44625535 on Build GRCh38
rs556190687 Chr.20: 44625675-44625675 on Build GRCh38
rs984176073 Chr.20: 44625721-44625721 on Build GRCh38
rs121908722 Chr.20: 44625580-44625580 on Build GRCh38
rs908247288 Chr.20: 44625724-44625724 on Build GRCh38
rs121908733 Chr.20: 44625602-44625602 on Build GRCh38
rs1015403283 Chr.20: 44625525-44625525 on Build GRCh38
rs867115027 Chr.20: 44625573-44625573 on Build GRCh38

View Full Product Details