Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55295436-55295705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC88A
Entrez Gene ID
55704
Gene Name
coiled-coil domain containing 88A
Gene Aliases
APE, GIRDIN, GIV, GRDN, HkRP1, KIAA1212
UniGene
Hs.292925
Gene Chromosome Location
Chr.2: 55287817-55419921 on Build GRCh38
Transcripts
NM_001135597.1 NM_001254943.1 NM_018084.4 XM_005264418.4 XM_005264421.2 XM_011532965.2 XM_011532966.2 XM_011532967.2 XM_011532968.2 XM_017004476.1 XM_017004477.1 XM_017004478.1 XM_017004479.1 XM_017004480.1 XM_017004481.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6622568 c.5390delT p.L1797fs*1 Chr.2: 55295674-55295674 on Build GRCh38
6622549 c.5447G>A p.R1816Q Chr.2: 55295698-55295698 on Build GRCh38
7416655 c.5524G>T p.D1842Y Chr.2: 55295621-55295621 on Build GRCh38
6222901 c.5478G>T p.K1826N Chr.2: 55295667-55295667 on Build GRCh38
6622550 c.5366G>A p.R1789Q Chr.2: 55295698-55295698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771428508 Chr.2: 55295544-55295544 on Build GRCh38
rs201745762 Chr.2: 55295589-55295589 on Build GRCh38
rs978832203 Chr.2: 55295564-55295564 on Build GRCh38
rs752753155 Chr.2: 55295538-55295538 on Build GRCh38
rs369578677 Chr.2: 55295587-55295587 on Build GRCh38
rs765383747 Chr.2: 55295659-55295659 on Build GRCh38
rs761231065 Chr.2: 55295569-55295569 on Build GRCh38
rs911710325 Chr.2: 55295609-55295609 on Build GRCh38
rs970000412 Chr.2: 55295561-55295561 on Build GRCh38
rs754649015 Chr.2: 55295542-55295542 on Build GRCh38
rs867681014 Chr.2: 55295653-55295653 on Build GRCh38
rs753681002 Chr.2: 55295533-55295533 on Build GRCh38
rs761334993 Chr.2: 55295492-55295492 on Build GRCh38
rs371854582 Chr.2: 55295555-55295555 on Build GRCh38
rs183431317 Chr.2: 55295662-55295662 on Build GRCh38
rs766334073 Chr.2: 55295523-55295523 on Build GRCh38
rs776781180 Chr.2: 55295554-55295554 on Build GRCh38
rs746247489 Chr.2: 55295681-55295681 on Build GRCh38
rs766387202 Chr.2: 55295614-55295614 on Build GRCh38
rs751610996 Chr.2: 55295664-55295664 on Build GRCh38
rs141590196 Chr.2: 55295661-55295661 on Build GRCh38
rs978528267 Chr.2: 55295603-55295603 on Build GRCh38
rs762583583 Chr.2: 55295483-55295483 on Build GRCh38
rs375237893 Chr.2: 55295556-55295556 on Build GRCh38
rs751061424 Chr.2: 55295490-55295491 on Build GRCh38
rs948456086 Chr.2: 55295502-55295502 on Build GRCh38
rs758438490 Chr.2: 55295540-55295540 on Build GRCh38
rs566753897 Chr.2: 55295567-55295567 on Build GRCh38
rs781629815 Chr.2: 55295678-55295678 on Build GRCh38
rs367582957 Chr.2: 55295651-55295651 on Build GRCh38
rs778266530 Chr.2: 55295542-55295542 on Build GRCh38
rs760556370 Chr.2: 55295512-55295512 on Build GRCh38
rs1052916861 Chr.2: 55295599-55295599 on Build GRCh38
rs781580277 Chr.2: 55295550-55295550 on Build GRCh38
rs750315586 Chr.2: 55295506-55295506 on Build GRCh38
rs546688615 Chr.2: 55295510-55295510 on Build GRCh38
rs1031558748 Chr.2: 55295622-55295622 on Build GRCh38
rs934730785 Chr.2: 55295595-55295595 on Build GRCh38
rs759324269 Chr.2: 55295655-55295655 on Build GRCh38
rs767591033 Chr.2: 55295495-55295495 on Build GRCh38
rs757813114 Chr.2: 55295673-55295673 on Build GRCh38
rs768326043 Chr.2: 55295486-55295486 on Build GRCh38
rs960360589 Chr.2: 55295530-55295530 on Build GRCh38
rs760761141 Chr.2: 55295598-55295598 on Build GRCh38
rs773784899 Chr.2: 55295490-55295490 on Build GRCh38
rs117795614 Chr.2: 55295464-55295464 on Build GRCh38

View Full Product Details