Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30663873-30664098 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP18
Entrez Gene ID
150290
Gene Name
dual specificity phosphatase 18
Gene Aliases
DSP18, DUSP20, LMWDSP20
UniGene
Hs.517544
Gene Chromosome Location
Chr.22: 30652051-30667890 on Build GRCh38
Transcripts
NM_001304794.1 NM_001304795.1 NM_001304796.1 NM_152511.4 XM_005261368.4 XM_006724148.3 XM_011529920.2 XM_011529921.2 XM_017028627.1 XM_017028628.1 NM_001318371.1

Gene Symbol
SLC35E4
Entrez Gene ID
339665
Gene Name
solute carrier family 35 member E4
Gene Aliases
-
UniGene
Hs.660384
Gene Chromosome Location
Chr.22: 30634148-30669016 on Build GRCh38
Transcripts
NM_001304794.1 NM_001304795.1 NM_001304796.1 NM_152511.4 XM_005261368.4 XM_006724148.3 XM_011529920.2 XM_011529921.2 XM_017028627.1 XM_017028628.1 NM_001318371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6270570 c.111C>A p.N37K Chr.22: 30663893-30663893 on Build GRCh38
444873 c.36C>T p.F12F Chr.22: 30663968-30663968 on Build GRCh38
725920 c.51C>T p.V17V Chr.22: 30663953-30663953 on Build GRCh38
4103478 c.102C>T p.A34A Chr.22: 30663902-30663902 on Build GRCh38
2937375 c.42G>T p.Q14H Chr.22: 30663962-30663962 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752463721 Chr.22: 30663977-30663977 on Build GRCh38
rs774890036 Chr.22: 30663950-30663950 on Build GRCh38
rs769987050 Chr.22: 30664034-30664034 on Build GRCh38
rs559815332 Chr.22: 30663980-30663980 on Build GRCh38
rs780400702 Chr.22: 30664013-30664014 on Build GRCh38
rs756663571 Chr.22: 30663982-30663982 on Build GRCh38
rs756544719 Chr.22: 30664013-30664013 on Build GRCh38
rs768855710 Chr.22: 30663949-30663949 on Build GRCh38
rs747567208 Chr.22: 30664009-30664009 on Build GRCh38
rs745547281 Chr.22: 30663942-30663942 on Build GRCh38
rs145095975 Chr.22: 30663987-30663987 on Build GRCh38
rs775755708 Chr.22: 30664035-30664035 on Build GRCh38
rs772636480 Chr.22: 30664003-30664003 on Build GRCh38
rs748563948 Chr.22: 30663956-30663956 on Build GRCh38
rs767397656 Chr.22: 30664051-30664051 on Build GRCh38
rs770873099 Chr.22: 30664010-30664010 on Build GRCh38
rs1050888093 Chr.22: 30663965-30663965 on Build GRCh38
rs754665481 Chr.22: 30663902-30663902 on Build GRCh38
rs200339726 Chr.22: 30663901-30663901 on Build GRCh38
rs1005887 Chr.22: 30664042-30664042 on Build GRCh38
rs752588621 Chr.22: 30663919-30663919 on Build GRCh38
rs377416793 Chr.22: 30663930-30663930 on Build GRCh38
rs760706123 Chr.22: 30664007-30664007 on Build GRCh38
rs766487745 Chr.22: 30663968-30663968 on Build GRCh38
rs1031088302 Chr.22: 30664012-30664012 on Build GRCh38
rs150392282 Chr.22: 30663971-30663971 on Build GRCh38
rs558283667 Chr.22: 30663984-30663984 on Build GRCh38
rs763887554 Chr.22: 30664040-30664040 on Build GRCh38
rs762668743 Chr.22: 30664038-30664038 on Build GRCh38
rs140453707 Chr.22: 30663966-30663966 on Build GRCh38
rs780641324 Chr.22: 30663938-30663938 on Build GRCh38
rs946735981 Chr.22: 30663920-30663920 on Build GRCh38
rs750448299 Chr.22: 30663912-30663912 on Build GRCh38
rs779902558 Chr.22: 30663990-30663990 on Build GRCh38
rs755797214 Chr.22: 30663989-30663989 on Build GRCh38
rs748513055 Chr.22: 30663998-30663998 on Build GRCh38
rs765479545 Chr.22: 30663976-30663976 on Build GRCh38
rs756858917 Chr.22: 30663937-30663937 on Build GRCh38
rs776739722 Chr.22: 30664016-30664016 on Build GRCh38
rs528509753 Chr.22: 30663981-30663981 on Build GRCh38
rs760671397 Chr.22: 30663967-30663967 on Build GRCh38
rs759580003 Chr.22: 30664028-30664028 on Build GRCh38
rs146808085 Chr.22: 30663964-30663964 on Build GRCh38
rs970357526 Chr.22: 30664008-30664008 on Build GRCh38
rs544877040 Chr.22: 30663942-30663943 on Build GRCh38
rs886318125 Chr.22: 30663921-30663921 on Build GRCh38
rs143675020 Chr.22: 30663933-30663933 on Build GRCh38
rs758246378 Chr.22: 30663978-30663978 on Build GRCh38

View Full Product Details