Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 49396496-49396737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf78
Entrez Gene ID
123970
Gene Name
chromosome 16 open reading frame 78
Gene Aliases
-
UniGene
Hs.125875
Gene Chromosome Location
Chr.16: 49373808-49399431 on Build GRCh38
Transcripts
NM_144602.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3818043 c.524G>A p.R175K Chr.16: 49396552-49396552 on Build GRCh38
4772660 c.559G>A p.E187K Chr.16: 49396587-49396587 on Build GRCh38
3691031 c.649C>T p.R217* Chr.16: 49396677-49396677 on Build GRCh38
6484405 c.485A>T p.Q162L Chr.16: 49396513-49396513 on Build GRCh38
4632125 c.512C>T p.A171V Chr.16: 49396540-49396540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770543456 Chr.16: 49396674-49396674 on Build GRCh38
rs772691046 Chr.16: 49396668-49396668 on Build GRCh38
rs756568543 Chr.16: 49396547-49396547 on Build GRCh38
rs144220022 Chr.16: 49396548-49396548 on Build GRCh38
rs72776789 Chr.16: 49396623-49396623 on Build GRCh38
rs368092475 Chr.16: 49396711-49396711 on Build GRCh38
rs201425004 Chr.16: 49396641-49396641 on Build GRCh38
rs759293691 Chr.16: 49396609-49396609 on Build GRCh38
rs746586351 Chr.16: 49396672-49396672 on Build GRCh38
rs377208417 Chr.16: 49396713-49396713 on Build GRCh38
rs764186843 Chr.16: 49396689-49396689 on Build GRCh38
rs148751828 Chr.16: 49396553-49396553 on Build GRCh38
rs374900464 Chr.16: 49396535-49396535 on Build GRCh38
rs749690623 Chr.16: 49396646-49396646 on Build GRCh38
rs757559961 Chr.16: 49396587-49396587 on Build GRCh38
rs752532359 Chr.16: 49396622-49396622 on Build GRCh38
rs940432278 Chr.16: 49396568-49396568 on Build GRCh38
rs1037336603 Chr.16: 49396573-49396573 on Build GRCh38
rs776198743 Chr.16: 49396677-49396677 on Build GRCh38
rs762032705 Chr.16: 49396691-49396691 on Build GRCh38
rs562874658 Chr.16: 49396679-49396679 on Build GRCh38
rs142163916 Chr.16: 49396608-49396608 on Build GRCh38
rs775361605 Chr.16: 49396681-49396681 on Build GRCh38
rs761269773 Chr.16: 49396543-49396543 on Build GRCh38
rs765158503 Chr.16: 49396566-49396566 on Build GRCh38
rs370873314 Chr.16: 49396576-49396576 on Build GRCh38
rs765201923 Chr.16: 49396614-49396614 on Build GRCh38
rs916520408 Chr.16: 49396531-49396531 on Build GRCh38
rs753777579 Chr.16: 49396692-49396692 on Build GRCh38
rs929751686 Chr.16: 49396698-49396698 on Build GRCh38
rs1046878164 Chr.16: 49396700-49396700 on Build GRCh38
rs772709408 Chr.16: 49396596-49396596 on Build GRCh38
rs752794574 Chr.16: 49396570-49396570 on Build GRCh38
rs949233644 Chr.16: 49396536-49396536 on Build GRCh38
rs760259078 Chr.16: 49396597-49396597 on Build GRCh38
rs766101249 Chr.16: 49396626-49396626 on Build GRCh38
rs754926704 Chr.16: 49396554-49396554 on Build GRCh38
rs147626517 Chr.16: 49396607-49396607 on Build GRCh38
rs756363297 Chr.16: 49396696-49396696 on Build GRCh38
rs767656781 Chr.16: 49396692-49396692 on Build GRCh38
rs767134665 Chr.16: 49396544-49396544 on Build GRCh38
rs147852211 Chr.16: 49396678-49396678 on Build GRCh38
rs747196284 Chr.16: 49396586-49396586 on Build GRCh38
rs531614691 Chr.16: 49396699-49396699 on Build GRCh38
rs1033098185 Chr.16: 49396659-49396659 on Build GRCh38
rs140378939 Chr.16: 49396590-49396590 on Build GRCh38
rs778024669 Chr.16: 49396578-49396578 on Build GRCh38
rs750571571 Chr.16: 49396695-49396695 on Build GRCh38
rs769161271 Chr.16: 49396656-49396656 on Build GRCh38
rs780498767 Chr.16: 49396645-49396645 on Build GRCh38
rs867806225 Chr.16: 49396635-49396635 on Build GRCh38
rs773899895 Chr.16: 49396537-49396537 on Build GRCh38
rs916481852 Chr.16: 49396704-49396704 on Build GRCh38
rs746276397 Chr.16: 49396589-49396589 on Build GRCh38
rs560343232 Chr.16: 49396638-49396638 on Build GRCh38
rs374603360 Chr.16: 49396591-49396591 on Build GRCh38
rs61605376 Chr.16: 49396543-49396554 on Build GRCh38
rs917017472 Chr.16: 49396683-49396683 on Build GRCh38
rs766093244 Chr.16: 49396552-49396552 on Build GRCh38
rs762926222 Chr.16: 49396682-49396682 on Build GRCh38
rs781337420 Chr.16: 49396632-49396632 on Build GRCh38
rs764203233 Chr.16: 49396624-49396624 on Build GRCh38

View Full Product Details