Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 18661894-18662129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
UniGene
Hs.741483
Gene Chromosome Location
Chr.9: 17906574-18910950 on Build GRCh38
Transcripts
NM_001040272.5 NM_052866.4 XM_011518063.2 XM_011518064.2 XM_011518067.1 XM_017015310.1 XM_017015311.1 XM_017015312.1 XM_017015313.1 XM_017015314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
158913 c.1012T>A p.C338S Chr.9: 18662000-18662000 on Build GRCh38
3745942 c.1069G>A p.D357N Chr.9: 18662057-18662057 on Build GRCh38
6991485 c.1002T>C p.A334A Chr.9: 18661990-18661990 on Build GRCh38
5101483 c.997G>A p.V333I Chr.9: 18661985-18661985 on Build GRCh38
3731041 c.1001C>T p.A334V Chr.9: 18661989-18661989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191918530 Chr.9: 18662036-18662036 on Build GRCh38
rs765624574 Chr.9: 18662067-18662067 on Build GRCh38
rs746762586 Chr.9: 18662085-18662085 on Build GRCh38
rs759899921 Chr.9: 18662062-18662062 on Build GRCh38
rs758162162 Chr.9: 18661929-18661929 on Build GRCh38
rs987282681 Chr.9: 18661988-18661988 on Build GRCh38
rs377552651 Chr.9: 18662081-18662081 on Build GRCh38
rs749267542 Chr.9: 18662015-18662015 on Build GRCh38
rs141659739 Chr.9: 18661997-18661997 on Build GRCh38
rs771820042 Chr.9: 18662035-18662035 on Build GRCh38
rs764202410 Chr.9: 18661985-18661985 on Build GRCh38
rs770603282 Chr.9: 18661942-18661942 on Build GRCh38
rs370253600 Chr.9: 18661974-18661974 on Build GRCh38
rs757559183 Chr.9: 18661994-18661994 on Build GRCh38
rs777583332 Chr.9: 18661950-18661950 on Build GRCh38
rs141627942 Chr.9: 18661963-18661963 on Build GRCh38
rs73433591 Chr.9: 18662014-18662014 on Build GRCh38
rs139287759 Chr.9: 18662020-18662020 on Build GRCh38
rs777106613 Chr.9: 18661981-18661981 on Build GRCh38
rs748749844 Chr.9: 18661967-18661967 on Build GRCh38
rs148178041 Chr.9: 18661951-18661951 on Build GRCh38
rs934136209 Chr.9: 18661998-18661998 on Build GRCh38
rs763501571 Chr.9: 18662088-18662088 on Build GRCh38
rs756246381 Chr.9: 18661964-18661964 on Build GRCh38
rs753230182 Chr.9: 18661984-18661984 on Build GRCh38
rs751760171 Chr.9: 18661990-18661990 on Build GRCh38
rs745610882 Chr.9: 18662013-18662013 on Build GRCh38
rs1038736765 Chr.9: 18662052-18662052 on Build GRCh38
rs142112150 Chr.9: 18661980-18661980 on Build GRCh38
rs775947988 Chr.9: 18662068-18662068 on Build GRCh38
rs150088929 Chr.9: 18661979-18661979 on Build GRCh38
rs747739409 Chr.9: 18662030-18662030 on Build GRCh38
rs775310518 Chr.9: 18662047-18662047 on Build GRCh38
rs986156040 Chr.9: 18662034-18662034 on Build GRCh38
rs973362151 Chr.9: 18661993-18661993 on Build GRCh38
rs376273773 Chr.9: 18661948-18661948 on Build GRCh38
rs537641249 Chr.9: 18661934-18661934 on Build GRCh38
rs768249524 Chr.9: 18662018-18662018 on Build GRCh38
rs758490070 Chr.9: 18661928-18661928 on Build GRCh38
rs574508977 Chr.9: 18661982-18661982 on Build GRCh38
rs774106541 Chr.9: 18661975-18661975 on Build GRCh38
rs558928988 Chr.9: 18661941-18661941 on Build GRCh38

View Full Product Details