Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232015467-232015714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIS3L2
Entrez Gene ID
129563
Gene Name
DIS3 like 3'-5' exoribonuclease 2
Gene Aliases
FAM6A, PRLMNS, hDIS3L2
UniGene
Hs.732236
Gene Chromosome Location
Chr.2: 231961583-232343968 on Build GRCh38
Transcripts
NM_001257281.1 NM_001257282.1 NM_152383.4 NR_046476.1 NR_046477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6990542 c.59C>G p.S20C Chr.2: 232015520-232015520 on Build GRCh38
5803515 c.210G>A p.Q70Q Chr.2: 232015671-232015671 on Build GRCh38
6010707 c.72T>G p.G24G Chr.2: 232015533-232015533 on Build GRCh38
6990545 c.59C>G p.S20C Chr.2: 232015520-232015520 on Build GRCh38
6448392 c.127C>T p.R43* Chr.2: 232015588-232015588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777447491 Chr.2: 232015657-232015657 on Build GRCh38
rs772722191 Chr.2: 232015554-232015554 on Build GRCh38
rs746627444 Chr.2: 232015672-232015672 on Build GRCh38
rs1000963300 Chr.2: 232015633-232015633 on Build GRCh38
rs774039141 Chr.2: 232015498-232015498 on Build GRCh38
rs1032683392 Chr.2: 232015550-232015550 on Build GRCh38
rs765044826 Chr.2: 232015496-232015496 on Build GRCh38
rs981586581 Chr.2: 232015634-232015634 on Build GRCh38
rs958950313 Chr.2: 232015652-232015652 on Build GRCh38
rs756250230 Chr.2: 232015536-232015536 on Build GRCh38
rs201117797 Chr.2: 232015681-232015681 on Build GRCh38
rs1004734586 Chr.2: 232015529-232015529 on Build GRCh38
rs758664325 Chr.2: 232015627-232015627 on Build GRCh38
rs754408232 Chr.2: 232015604-232015604 on Build GRCh38
rs776585118 Chr.2: 232015562-232015562 on Build GRCh38
rs762653147 Chr.2: 232015588-232015588 on Build GRCh38
rs372201663 Chr.2: 232015517-232015517 on Build GRCh38
rs767500228 Chr.2: 232015533-232015533 on Build GRCh38
rs770400666 Chr.2: 232015561-232015561 on Build GRCh38
rs886288172 Chr.2: 232015528-232015528 on Build GRCh38
rs755318575 Chr.2: 232015544-232015544 on Build GRCh38
rs376019404 Chr.2: 232015670-232015670 on Build GRCh38
rs866879001 Chr.2: 232015658-232015658 on Build GRCh38
rs901797759 Chr.2: 232015553-232015553 on Build GRCh38
rs530571890 Chr.2: 232015621-232015621 on Build GRCh38
rs968496794 Chr.2: 232015626-232015626 on Build GRCh38
rs759322216 Chr.2: 232015530-232015531 on Build GRCh38
rs756293168 Chr.2: 232015597-232015597 on Build GRCh38
rs779281100 Chr.2: 232015620-232015620 on Build GRCh38
rs764957786 Chr.2: 232015563-232015563 on Build GRCh38
rs755444049 Chr.2: 232015619-232015619 on Build GRCh38
rs779277888 Chr.2: 232015546-232015546 on Build GRCh38
rs375960593 Chr.2: 232015519-232015519 on Build GRCh38
rs750642629 Chr.2: 232015591-232015591 on Build GRCh38
rs368468992 Chr.2: 232015495-232015495 on Build GRCh38
rs1031144721 Chr.2: 232015636-232015636 on Build GRCh38
rs750597953 Chr.2: 232015534-232015534 on Build GRCh38
rs749771898 Chr.2: 232015543-232015543 on Build GRCh38
rs183463487 Chr.2: 232015525-232015525 on Build GRCh38
rs746567735 Chr.2: 232015558-232015558 on Build GRCh38
rs766516846 Chr.2: 232015602-232015602 on Build GRCh38
rs780747005 Chr.2: 232015540-232015540 on Build GRCh38
rs369080386 Chr.2: 232015547-232015547 on Build GRCh38
rs371477071 Chr.2: 232015569-232015569 on Build GRCh38
rs929991353 Chr.2: 232015682-232015683 on Build GRCh38
rs767837521 Chr.2: 232015589-232015589 on Build GRCh38

View Full Product Details