Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43187874-43188147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL9
Entrez Gene ID
23113
Gene Name
cullin 9
Gene Aliases
H7AP1, PARC
UniGene
Hs.485434
Gene Chromosome Location
Chr.6: 43182170-43224587 on Build GRCh38
Transcripts
NM_015089.3 XM_011514422.1 XM_011514423.2 XM_011514424.1 XM_011514425.1 XM_011514426.1 XM_011514427.1 XM_011514428.1 XM_011514429.1 XM_011514430.1 XM_017010589.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6836838 c.1892A>G p.E631G Chr.6: 43188023-43188023 on Build GRCh38
7079905 c.1846G>T p.A616S Chr.6: 43187977-43187977 on Build GRCh38
307728 c.1781C>T p.P594L Chr.6: 43187912-43187912 on Build GRCh38
4169931 c.1849G>A p.E617K Chr.6: 43187980-43187980 on Build GRCh38
451512 c.1848C>T p.A616A Chr.6: 43187979-43187979 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142314571 Chr.6: 43187979-43187979 on Build GRCh38
rs576021659 Chr.6: 43188088-43188088 on Build GRCh38
rs755371620 Chr.6: 43188115-43188115 on Build GRCh38
rs755530932 Chr.6: 43188001-43188001 on Build GRCh38
rs375220618 Chr.6: 43188021-43188021 on Build GRCh38
rs753044337 Chr.6: 43188125-43188125 on Build GRCh38
rs745960644 Chr.6: 43188101-43188101 on Build GRCh38
rs779104958 Chr.6: 43188015-43188015 on Build GRCh38
rs138491396 Chr.6: 43188000-43188000 on Build GRCh38
rs556157362 Chr.6: 43188085-43188085 on Build GRCh38
rs145911635 Chr.6: 43187980-43187980 on Build GRCh38
rs764896459 Chr.6: 43187948-43187948 on Build GRCh38
rs758826559 Chr.6: 43188126-43188126 on Build GRCh38
rs925658083 Chr.6: 43188094-43188094 on Build GRCh38
rs776799047 Chr.6: 43188029-43188029 on Build GRCh38
rs201640275 Chr.6: 43188022-43188022 on Build GRCh38
rs769778026 Chr.6: 43188054-43188054 on Build GRCh38
rs140466677 Chr.6: 43188026-43188026 on Build GRCh38
rs145896671 Chr.6: 43188082-43188082 on Build GRCh38
rs745757434 Chr.6: 43188031-43188031 on Build GRCh38
rs145277223 Chr.6: 43187899-43187899 on Build GRCh38
rs772309828 Chr.6: 43187903-43187903 on Build GRCh38
rs760754012 Chr.6: 43187913-43187913 on Build GRCh38
rs781308070 Chr.6: 43188069-43188069 on Build GRCh38
rs762631160 Chr.6: 43187958-43187958 on Build GRCh38
rs773208772 Chr.6: 43187906-43187906 on Build GRCh38
rs751103085 Chr.6: 43187962-43187962 on Build GRCh38
rs375887998 Chr.6: 43187936-43187936 on Build GRCh38
rs775370901 Chr.6: 43188066-43188066 on Build GRCh38
rs752572922 Chr.6: 43187950-43187950 on Build GRCh38
rs201588709 Chr.6: 43187930-43187930 on Build GRCh38
rs774188388 Chr.6: 43188079-43188079 on Build GRCh38
rs759356800 Chr.6: 43187931-43187931 on Build GRCh38
rs770883961 Chr.6: 43187916-43187916 on Build GRCh38
rs368909436 Chr.6: 43187961-43187961 on Build GRCh38
rs770952969 Chr.6: 43188028-43188028 on Build GRCh38
rs149158952 Chr.6: 43187911-43187911 on Build GRCh38

View Full Product Details