Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74623463-74623736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD300E
Entrez Gene ID
342510
Gene Name
CD300e molecule
Gene Aliases
CD300LE, CLM-2, CLM2, CMRF35-A5, IREM-2, IREM2, PIgR-2, PIgR2
UniGene
Hs.158954
Gene Chromosome Location
Chr.17: 74609883-74623958 on Build GRCh38
Transcripts
NM_181449.2 XM_017024575.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
437326 c.56C>T p.P19L Chr.17: 74623689-74623689 on Build GRCh38
5451458 c.10C>A p.L4I Chr.17: 74623612-74623612 on Build GRCh38
3362347 c.13C>T p.P5S Chr.17: 74623609-74623609 on Build GRCh38
2977765 c.163+5G>A p.? Chr.17: 74623577-74623577 on Build GRCh38
6147995 c.82G>T p.G28* Chr.17: 74623663-74623663 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746484713 Chr.17: 74623694-74623694 on Build GRCh38
rs757854780 Chr.17: 74623689-74623689 on Build GRCh38
rs771424801 Chr.17: 74623553-74623553 on Build GRCh38
rs780748011 Chr.17: 74623503-74623503 on Build GRCh38
rs768515171 Chr.17: 74623583-74623583 on Build GRCh38
rs775173098 Chr.17: 74623567-74623567 on Build GRCh38
rs200851226 Chr.17: 74623709-74623709 on Build GRCh38
rs41458047 Chr.17: 74623656-74623656 on Build GRCh38
rs779588520 Chr.17: 74623642-74623642 on Build GRCh38
rs375091102 Chr.17: 74623643-74623643 on Build GRCh38
rs1027672241 Chr.17: 74623505-74623505 on Build GRCh38
rs973483842 Chr.17: 74623496-74623496 on Build GRCh38
rs753043255 Chr.17: 74623534-74623534 on Build GRCh38
rs760947974 Chr.17: 74623677-74623677 on Build GRCh38
rs115257540 Chr.17: 74623488-74623488 on Build GRCh38
rs1002669754 Chr.17: 74623510-74623510 on Build GRCh38
rs753416006 Chr.17: 74623539-74623540 on Build GRCh38
rs749736465 Chr.17: 74623542-74623542 on Build GRCh38
rs767663532 Chr.17: 74623615-74623615 on Build GRCh38
rs778178588 Chr.17: 74623620-74623620 on Build GRCh38
rs778454271 Chr.17: 74623554-74623554 on Build GRCh38
rs116269002 Chr.17: 74623612-74623612 on Build GRCh38
rs779678436 Chr.17: 74623693-74623693 on Build GRCh38
rs771662321 Chr.17: 74623565-74623565 on Build GRCh38
rs754243021 Chr.17: 74623683-74623683 on Build GRCh38
rs779362248 Chr.17: 74623632-74623632 on Build GRCh38
rs752913255 Chr.17: 74623616-74623616 on Build GRCh38
rs771564789 Chr.17: 74623637-74623637 on Build GRCh38
rs778274305 Chr.17: 74623540-74623540 on Build GRCh38
rs757578072 Chr.17: 74623684-74623684 on Build GRCh38
rs200133927 Chr.17: 74623622-74623622 on Build GRCh38
rs745404618 Chr.17: 74623634-74623634 on Build GRCh38
rs529798930 Chr.17: 74623598-74623598 on Build GRCh38
rs373450409 Chr.17: 74623563-74623563 on Build GRCh38
rs756459607 Chr.17: 74623539-74623539 on Build GRCh38
rs759630438 Chr.17: 74623613-74623613 on Build GRCh38
rs767575109 Chr.17: 74623666-74623666 on Build GRCh38
rs370602651 Chr.17: 74623619-74623619 on Build GRCh38
rs768417203 Chr.17: 74623648-74623648 on Build GRCh38
rs35241963 Chr.17: 74623581-74623581 on Build GRCh38
rs776623350 Chr.17: 74623654-74623654 on Build GRCh38
rs142035923 Chr.17: 74623623-74623623 on Build GRCh38
rs754723041 Chr.17: 74623705-74623705 on Build GRCh38
rs776712708 Chr.17: 74623591-74623591 on Build GRCh38
rs745421874 Chr.17: 74623519-74623519 on Build GRCh38
rs761808938 Chr.17: 74623662-74623662 on Build GRCh38
rs867573915 Chr.17: 74623594-74623594 on Build GRCh38
rs752865488 Chr.17: 74623674-74623674 on Build GRCh38
rs560846024 Chr.17: 74623597-74623597 on Build GRCh38
rs572219391 Chr.17: 74623688-74623688 on Build GRCh38
rs955733694 Chr.17: 74623638-74623638 on Build GRCh38
rs767397737 Chr.17: 74623681-74623681 on Build GRCh38
rs189565267 Chr.17: 74623579-74623579 on Build GRCh38

View Full Product Details