Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 111734971-111735244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK4
Entrez Gene ID
9732
Gene Name
dedicator of cytokinesis 4
Gene Aliases
-
UniGene
Hs.654652
Gene Chromosome Location
Chr.7: 111726108-112206407 on Build GRCh38
Transcripts
NM_014705.3 XM_006716188.2 XM_006716189.2 XM_017012819.1 XM_017012820.1 XM_017012821.1 XM_017012822.1 XM_017012823.1 XM_017012824.1 XM_017012825.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5573259 c.5392+1G>C p.? Chr.7: 111735053-111735053 on Build GRCh38
6301569 c.5303G>A p.W1768* Chr.7: 111735143-111735143 on Build GRCh38
5894862 c.2011C>T p.P671S Chr.7: 111735084-111735084 on Build GRCh38
5599432 c.1943C>T p.P648L Chr.7: 111735152-111735152 on Build GRCh38
6272327 c.1940C>A p.T647N Chr.7: 111735155-111735155 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763040540 Chr.7: 111735139-111735139 on Build GRCh38
rs764347403 Chr.7: 111735032-111735032 on Build GRCh38
rs139961687 Chr.7: 111735106-111735106 on Build GRCh38
rs373891157 Chr.7: 111735101-111735101 on Build GRCh38
rs375916291 Chr.7: 111735182-111735182 on Build GRCh38
rs553835973 Chr.7: 111735070-111735070 on Build GRCh38
rs372909552 Chr.7: 111735198-111735198 on Build GRCh38
rs564121451 Chr.7: 111735089-111735089 on Build GRCh38
rs778520958 Chr.7: 111735074-111735074 on Build GRCh38
rs199565386 Chr.7: 111735090-111735090 on Build GRCh38
rs746809269 Chr.7: 111735084-111735084 on Build GRCh38
rs201460298 Chr.7: 111735081-111735081 on Build GRCh38
rs762780907 Chr.7: 111735136-111735136 on Build GRCh38
rs912636511 Chr.7: 111735184-111735184 on Build GRCh38
rs190576418 Chr.7: 111735018-111735018 on Build GRCh38
rs752859267 Chr.7: 111735010-111735010 on Build GRCh38
rs769731508 Chr.7: 111735107-111735107 on Build GRCh38
rs571343796 Chr.7: 111735043-111735044 on Build GRCh38
rs921393591 Chr.7: 111735158-111735158 on Build GRCh38
rs757801430 Chr.7: 111735043-111735043 on Build GRCh38
rs768057665 Chr.7: 111735160-111735160 on Build GRCh38
rs764412171 Chr.7: 111735149-111735149 on Build GRCh38
rs958237349 Chr.7: 111735211-111735211 on Build GRCh38
rs781766350 Chr.7: 111735057-111735057 on Build GRCh38
rs764598793 Chr.7: 111735191-111735191 on Build GRCh38
rs879204757 Chr.7: 111735056-111735056 on Build GRCh38
rs747730810 Chr.7: 111735080-111735080 on Build GRCh38
rs199968453 Chr.7: 111735201-111735201 on Build GRCh38
rs759412878 Chr.7: 111735199-111735199 on Build GRCh38
rs1013615451 Chr.7: 111735100-111735100 on Build GRCh38
rs763763027 Chr.7: 111735029-111735032 on Build GRCh38
rs750926729 Chr.7: 111735180-111735180 on Build GRCh38
rs774792111 Chr.7: 111735155-111735155 on Build GRCh38
rs371352582 Chr.7: 111735115-111735115 on Build GRCh38
rs751998356 Chr.7: 111735036-111735036 on Build GRCh38
rs368222477 Chr.7: 111735072-111735072 on Build GRCh38
rs954256261 Chr.7: 111735176-111735176 on Build GRCh38
rs757922165 Chr.7: 111735196-111735196 on Build GRCh38
rs368636490 Chr.7: 111735193-111735193 on Build GRCh38
rs762284440 Chr.7: 111735157-111735157 on Build GRCh38
rs545933889 Chr.7: 111735083-111735083 on Build GRCh38

View Full Product Details