Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80108095-80108368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAA
Entrez Gene ID
2548
Gene Name
glucosidase alpha, acid
Gene Aliases
LYAG
UniGene
Hs.1437
Gene Chromosome Location
Chr.17: 80101526-80119882 on Build GRCh38
Transcripts
NM_000152.4 NM_001079803.2 NM_001079804.2 XM_005257193.2 XM_005257194.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4623686 c.1012G>T p.D338Y Chr.17: 80108346-80108346 on Build GRCh38
1252918 c.996G>A p.S332S Chr.17: 80108330-80108330 on Build GRCh38
4272958 c.1001G>A p.G334D Chr.17: 80108335-80108335 on Build GRCh38
6668353 c.971C>T p.P324L Chr.17: 80108305-80108305 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1041647959 Chr.17: 80108120-80108120 on Build GRCh38
rs779266179 Chr.17: 80108330-80108330 on Build GRCh38
rs775384900 Chr.17: 80108345-80108345 on Build GRCh38
rs895612400 Chr.17: 80108295-80108295 on Build GRCh38
rs778227292 Chr.17: 80108321-80108321 on Build GRCh38
rs750030887 Chr.17: 80108305-80108305 on Build GRCh38
rs757608620 Chr.17: 80108329-80108329 on Build GRCh38
rs756562462 Chr.17: 80108313-80108313 on Build GRCh38
rs202095215 Chr.17: 80108337-80108337 on Build GRCh38
rs778172657 Chr.17: 80108292-80108292 on Build GRCh38
rs962925012 Chr.17: 80108231-80108231 on Build GRCh38
rs539097550 Chr.17: 80108226-80108226 on Build GRCh38
rs2241888 Chr.17: 80108183-80108183 on Build GRCh38
rs571369729 Chr.17: 80108207-80108207 on Build GRCh38
rs765190225 Chr.17: 80108278-80108278 on Build GRCh38
rs983718782 Chr.17: 80108154-80108154 on Build GRCh38
rs1012323041 Chr.17: 80108218-80108218 on Build GRCh38
rs528420839 Chr.17: 80108192-80108192 on Build GRCh38
rs763445243 Chr.17: 80108306-80108306 on Build GRCh38
rs201713593 Chr.17: 80108259-80108259 on Build GRCh38
rs959357014 Chr.17: 80108217-80108217 on Build GRCh38
rs2241887 Chr.17: 80108206-80108206 on Build GRCh38
rs1044137447 Chr.17: 80108245-80108245 on Build GRCh38
rs749831587 Chr.17: 80108327-80108327 on Build GRCh38
rs955780209 Chr.17: 80108347-80108347 on Build GRCh38
rs747100906 Chr.17: 80108198-80108198 on Build GRCh38
rs994615279 Chr.17: 80108342-80108342 on Build GRCh38
rs905633601 Chr.17: 80108308-80108308 on Build GRCh38
rs369149976 Chr.17: 80108282-80108282 on Build GRCh38
rs761670883 Chr.17: 80108262-80108262 on Build GRCh38
rs1034947613 Chr.17: 80108209-80108209 on Build GRCh38
rs1010471584 Chr.17: 80108118-80108118 on Build GRCh38
rs892992366 Chr.17: 80108117-80108117 on Build GRCh38
rs886042960 Chr.17: 80108334-80108334 on Build GRCh38
rs372714540 Chr.17: 80108248-80108248 on Build GRCh38
rs950923867 Chr.17: 80108128-80108128 on Build GRCh38
rs772371937 Chr.17: 80108341-80108341 on Build GRCh38
rs730880022 Chr.17: 80108338-80108338 on Build GRCh38
rs868423260 Chr.17: 80108236-80108236 on Build GRCh38

View Full Product Details