Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100683093-100683332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIGYF1
Entrez Gene ID
64599
Gene Name
GRB10 interacting GYF protein 1
Gene Aliases
GYF1, PERQ1
UniGene
Hs.414396
Gene Chromosome Location
Chr.7: 100679507-100694282 on Build GRCh38
Transcripts
NM_022574.4 XM_005250532.1 XM_011516472.1 XM_011516477.1 XM_017012526.1 XM_017012527.1 XM_017012528.1 XM_017012529.1 XM_017012530.1 XM_017012531.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1312475 c.2280C>G p.L760L Chr.7: 100683144-100683144 on Build GRCh38
6670900 c.1328A>G p.E443G Chr.7: 100683326-100683326 on Build GRCh38
54536 c.2178G>A p.L726L Chr.7: 100683319-100683319 on Build GRCh38
7089575 c.2256C>A p.P752P Chr.7: 100683168-100683168 on Build GRCh38
3080715 c.2240C>T p.A747V Chr.7: 100683184-100683184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766718677 Chr.7: 100683216-100683216 on Build GRCh38
rs757429798 Chr.7: 100683310-100683310 on Build GRCh38
rs541882700 Chr.7: 100683135-100683135 on Build GRCh38
rs943948097 Chr.7: 100683175-100683175 on Build GRCh38
rs372612503 Chr.7: 100683245-100683245 on Build GRCh38
rs747779027 Chr.7: 100683271-100683271 on Build GRCh38
rs945356104 Chr.7: 100683207-100683207 on Build GRCh38
rs977792634 Chr.7: 100683213-100683213 on Build GRCh38
rs143878382 Chr.7: 100683304-100683304 on Build GRCh38
rs148635376 Chr.7: 100683262-100683262 on Build GRCh38
rs759808299 Chr.7: 100683275-100683275 on Build GRCh38
rs750871793 Chr.7: 100683236-100683236 on Build GRCh38
rs771883212 Chr.7: 100683253-100683253 on Build GRCh38
rs768006572 Chr.7: 100683234-100683235 on Build GRCh38
rs745766057 Chr.7: 100683173-100683173 on Build GRCh38
rs761593309 Chr.7: 100683205-100683205 on Build GRCh38
rs221793 Chr.7: 100683273-100683273 on Build GRCh38
rs376344485 Chr.7: 100683261-100683261 on Build GRCh38
rs756254194 Chr.7: 100683154-100683154 on Build GRCh38
rs773063004 Chr.7: 100683254-100683254 on Build GRCh38
rs568247705 Chr.7: 100683311-100683311 on Build GRCh38
rs772985821 Chr.7: 100683274-100683274 on Build GRCh38
rs563987336 Chr.7: 100683178-100683178 on Build GRCh38
rs939474720 Chr.7: 100683282-100683282 on Build GRCh38
rs778603866 Chr.7: 100683172-100683172 on Build GRCh38
rs757629443 Chr.7: 100683185-100683186 on Build GRCh38
rs755046743 Chr.7: 100683281-100683284 on Build GRCh38
rs767907287 Chr.7: 100683227-100683227 on Build GRCh38
rs749362923 Chr.7: 100683270-100683271 on Build GRCh38
rs777957103 Chr.7: 100683161-100683161 on Build GRCh38
rs760024412 Chr.7: 100683226-100683226 on Build GRCh38
rs911032111 Chr.7: 100683144-100683144 on Build GRCh38
rs746509851 Chr.7: 100683192-100683192 on Build GRCh38
rs967726034 Chr.7: 100683195-100683195 on Build GRCh38
rs767972301 Chr.7: 100683276-100683276 on Build GRCh38
rs1012289943 Chr.7: 100683184-100683184 on Build GRCh38
rs753925214 Chr.7: 100683307-100683307 on Build GRCh38
rs369031943 Chr.7: 100683241-100683241 on Build GRCh38
rs775943625 Chr.7: 100683284-100683284 on Build GRCh38
rs752141510 Chr.7: 100683167-100683168 on Build GRCh38
rs201771840 Chr.7: 100683252-100683252 on Build GRCh38
rs374629274 Chr.7: 100683269-100683269 on Build GRCh38
rs774593198 Chr.7: 100683222-100683222 on Build GRCh38
rs749558916 Chr.7: 100683168-100683168 on Build GRCh38
rs1021907570 Chr.7: 100683185-100683185 on Build GRCh38
rs752780672 Chr.7: 100683153-100683153 on Build GRCh38
rs776396510 Chr.7: 100683204-100683204 on Build GRCh38
rs761201481 Chr.7: 100683300-100683300 on Build GRCh38
rs774995550 Chr.7: 100683183-100683183 on Build GRCh38
rs752069062 Chr.7: 100683148-100683148 on Build GRCh38
rs531083829 Chr.7: 100683198-100683198 on Build GRCh38
rs377347579 Chr.7: 100683266-100683266 on Build GRCh38
rs770728430 Chr.7: 100683169-100683169 on Build GRCh38
rs756162193 Chr.7: 100683236-100683236 on Build GRCh38
rs756351073 Chr.7: 100683243-100683244 on Build GRCh38
rs780029270 Chr.7: 100683264-100683264 on Build GRCh38
rs781275978 Chr.7: 100683150-100683150 on Build GRCh38
rs573370693 Chr.7: 100683269-100683270 on Build GRCh38
rs753259209 Chr.7: 100683230-100683230 on Build GRCh38
rs936730908 Chr.7: 100683235-100683235 on Build GRCh38

View Full Product Details