Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 143439974-143440207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB1
Entrez Gene ID
2549
Gene Name
GRB2 associated binding protein 1
Gene Aliases
-
UniGene
Hs.618456
Gene Chromosome Location
Chr.4: 143336830-143474565 on Build GRCh38
Transcripts
NM_002039.3 NM_207123.2 XM_006714167.3 XM_006714168.3 XM_017007966.1 XM_017007967.1 XM_017007968.1 XM_017007969.1 XM_017007970.1 XM_017007971.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1427314 c.1327C>T p.L443L Chr.4: 143440124-143440124 on Build GRCh38
3600567 c.1347A>G p.P449P Chr.4: 143440144-143440144 on Build GRCh38
5346830 c.1367C>A p.P456Q Chr.4: 143440164-143440164 on Build GRCh38
4451573 c.1292A>G p.N431S Chr.4: 143440089-143440089 on Build GRCh38
5554233 c.1387T>G p.F463V Chr.4: 143440184-143440184 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968151837 Chr.4: 143440121-143440121 on Build GRCh38
rs201629079 Chr.4: 143440138-143440138 on Build GRCh38
rs765455903 Chr.4: 143440129-143440129 on Build GRCh38
rs112832809 Chr.4: 143440037-143440037 on Build GRCh38
rs28925906 Chr.4: 143440013-143440013 on Build GRCh38
rs866237568 Chr.4: 143440131-143440131 on Build GRCh38
rs916647203 Chr.4: 143440120-143440120 on Build GRCh38
rs774197436 Chr.4: 143440080-143440080 on Build GRCh38
rs760222411 Chr.4: 143440104-143440104 on Build GRCh38
rs958774873 Chr.4: 143440094-143440094 on Build GRCh38
rs770970128 Chr.4: 143440165-143440167 on Build GRCh38
rs776276159 Chr.4: 143440089-143440089 on Build GRCh38
rs758949394 Chr.4: 143440029-143440029 on Build GRCh38
rs779958695 Chr.4: 143440031-143440031 on Build GRCh38
rs1015034652 Chr.4: 143440030-143440030 on Build GRCh38
rs780650312 Chr.4: 143440144-143440144 on Build GRCh38
rs755165549 Chr.4: 143440151-143440151 on Build GRCh38
rs749080077 Chr.4: 143440067-143440067 on Build GRCh38
rs756917787 Chr.4: 143440127-143440127 on Build GRCh38
rs763734073 Chr.4: 143440112-143440112 on Build GRCh38
rs1010123836 Chr.4: 143440163-143440163 on Build GRCh38
rs767509569 Chr.4: 143440098-143440098 on Build GRCh38
rs534783471 Chr.4: 143440139-143440139 on Build GRCh38
rs991357606 Chr.4: 143440117-143440117 on Build GRCh38
rs376615337 Chr.4: 143440103-143440103 on Build GRCh38
rs950235452 Chr.4: 143440017-143440017 on Build GRCh38
rs142207353 Chr.4: 143440171-143440171 on Build GRCh38
rs370486761 Chr.4: 143440039-143440039 on Build GRCh38
rs776512236 Chr.4: 143440099-143440099 on Build GRCh38
rs751683961 Chr.4: 143440145-143440145 on Build GRCh38
rs753549802 Chr.4: 143440124-143440124 on Build GRCh38
rs369607803 Chr.4: 143440065-143440065 on Build GRCh38
rs748312293 Chr.4: 143440168-143440168 on Build GRCh38
rs770570752 Chr.4: 143440076-143440076 on Build GRCh38
rs778520256 Chr.4: 143440178-143440178 on Build GRCh38
rs369383480 Chr.4: 143440167-143440167 on Build GRCh38

View Full Product Details