Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 101828380-101828605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNIP2
Entrez Gene ID
30819
Gene Name
potassium voltage-gated channel interacting protein 2
Gene Aliases
KCHIP2
UniGene
Hs.97044
Gene Chromosome Location
Chr.10: 101825971-101843920 on Build GRCh38
Transcripts
NM_014591.4 NM_173191.2 NM_173192.2 NM_173193.2 NM_173194.2 NM_173195.2 NM_173197.2 XM_005269729.2 XM_005269730.2 XM_006717812.2 XM_011539731.2 XM_017016161.1 NR_045118.1

Gene Symbol
KCNIP2-AS1
Entrez Gene ID
100289509
Gene Name
KCNIP2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.568881
Gene Chromosome Location
Chr.10: 101819068-101828889 on Build GRCh38
Transcripts
NM_014591.4 NM_173191.2 NM_173192.2 NM_173193.2 NM_173194.2 NM_173195.2 NM_173197.2 XM_005269729.2 XM_005269730.2 XM_006717812.2 XM_011539731.2 XM_017016161.1 NR_045118.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6323341 c.330C>A p.V110V Chr.10: 101828398-101828398 on Build GRCh38
5821418 c.338A>G p.H113R Chr.10: 101828411-101828411 on Build GRCh38
5998980 c.354+6C>T p.? Chr.10: 101828383-101828383 on Build GRCh38
6323339 c.351C>A p.V117V Chr.10: 101828398-101828398 on Build GRCh38
5998978 c.534+6C>T p.? Chr.10: 101828383-101828383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765535056 Chr.10: 101828499-101828519 on Build GRCh38
rs760678229 Chr.10: 101828508-101828508 on Build GRCh38
rs774742355 Chr.10: 101828464-101828464 on Build GRCh38
rs769948374 Chr.10: 101828407-101828407 on Build GRCh38
rs961097554 Chr.10: 101828572-101828572 on Build GRCh38
rs764979702 Chr.10: 101828482-101828482 on Build GRCh38
rs1013668220 Chr.10: 101828520-101828520 on Build GRCh38
rs1055613832 Chr.10: 101828524-101828524 on Build GRCh38
rs770578421 Chr.10: 101828507-101828510 on Build GRCh38
rs762341879 Chr.10: 101828467-101828467 on Build GRCh38
rs765810060 Chr.10: 101828469-101828469 on Build GRCh38
rs1024619397 Chr.10: 101828541-101828541 on Build GRCh38
rs1046750310 Chr.10: 101828576-101828576 on Build GRCh38
rs753330095 Chr.10: 101828473-101828473 on Build GRCh38
rs759321585 Chr.10: 101828475-101828475 on Build GRCh38
rs763269682 Chr.10: 101828422-101828422 on Build GRCh38
rs754989333 Chr.10: 101828489-101828489 on Build GRCh38
rs201180498 Chr.10: 101828401-101828401 on Build GRCh38
rs199790708 Chr.10: 101828419-101828419 on Build GRCh38
rs769251762 Chr.10: 101828460-101828460 on Build GRCh38

View Full Product Details