Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11658548-11658781 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO44
Entrez Gene ID
93611
Gene Name
F-box protein 44
Gene Aliases
FBG3, FBX30, FBX6A, Fbx44, Fbxo6a
UniGene
Hs.556006
Gene Chromosome Location
Chr.1: 11654375-11663327 on Build GRCh38
Transcripts
NM_001014765.1 NM_001304790.1 NM_001304791.1 NM_033182.5 NM_183412.2 NM_183413.2 NR_130909.1 XM_005263535.1 XM_005263536.4 XM_005263537.1 XM_006711043.2 XM_006711045.2 XM_011542435.1 XM_017002842.1 XM_017002843.1 XM_017002844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7116434 c.450G>A p.L150L Chr.1: 11658590-11658590 on Build GRCh38
6275695 c.525G>T p.Q175H Chr.1: 11658772-11658772 on Build GRCh38
2178547 c.429C>T p.A143A Chr.1: 11658569-11658569 on Build GRCh38
6388621 c.439T>A p.W147R Chr.1: 11658579-11658579 on Build GRCh38
3471908 c.492C>T p.F164F Chr.1: 11658739-11658739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764944524 Chr.1: 11658688-11658688 on Build GRCh38
rs142346784 Chr.1: 11658740-11658740 on Build GRCh38
rs200779554 Chr.1: 11658643-11658643 on Build GRCh38
rs759439362 Chr.1: 11658726-11658726 on Build GRCh38
rs201882677 Chr.1: 11658698-11658698 on Build GRCh38
rs955379156 Chr.1: 11658590-11658590 on Build GRCh38
rs565261668 Chr.1: 11658703-11658703 on Build GRCh38
rs780557168 Chr.1: 11658755-11658755 on Build GRCh38
rs199660624 Chr.1: 11658569-11658569 on Build GRCh38
rs547985058 Chr.1: 11658610-11658610 on Build GRCh38
rs951119532 Chr.1: 11658680-11658680 on Build GRCh38
rs74054208 Chr.1: 11658704-11658704 on Build GRCh38
rs144418573 Chr.1: 11658614-11658614 on Build GRCh38
rs773242935 Chr.1: 11658706-11658706 on Build GRCh38
rs753718849 Chr.1: 11658656-11658656 on Build GRCh38
rs572795365 Chr.1: 11658683-11658683 on Build GRCh38
rs760967785 Chr.1: 11658711-11658711 on Build GRCh38
rs543294017 Chr.1: 11658685-11658685 on Build GRCh38
rs375259743 Chr.1: 11658589-11658589 on Build GRCh38
rs751112563 Chr.1: 11658745-11658745 on Build GRCh38
rs878970006 Chr.1: 11658744-11658744 on Build GRCh38
rs375500495 Chr.1: 11658667-11658667 on Build GRCh38
rs140912321 Chr.1: 11658618-11658618 on Build GRCh38
rs200078448 Chr.1: 11658686-11658686 on Build GRCh38
rs752667911 Chr.1: 11658743-11658755 on Build GRCh38
rs765282099 Chr.1: 11658708-11658708 on Build GRCh38
rs369457346 Chr.1: 11658712-11658712 on Build GRCh38
rs376877549 Chr.1: 11658675-11658675 on Build GRCh38
rs754966515 Chr.1: 11658665-11658665 on Build GRCh38
rs749723056 Chr.1: 11658757-11658757 on Build GRCh38
rs766203545 Chr.1: 11658653-11658653 on Build GRCh38
rs756987946 Chr.1: 11658751-11658751 on Build GRCh38
rs372518301 Chr.1: 11658758-11658758 on Build GRCh38
rs536114166 Chr.1: 11658604-11658604 on Build GRCh38
rs552972903 Chr.1: 11658682-11658682 on Build GRCh38
rs780575796 Chr.1: 11658692-11658692 on Build GRCh38
rs534641827 Chr.1: 11658679-11658679 on Build GRCh38
rs985356444 Chr.1: 11658736-11658736 on Build GRCh38
rs774626576 Chr.1: 11658620-11658620 on Build GRCh38
rs368147142 Chr.1: 11658607-11658607 on Build GRCh38
rs112449604 Chr.1: 11658674-11658674 on Build GRCh38
rs369440151 Chr.1: 11658673-11658673 on Build GRCh38
rs759623811 Chr.1: 11658604-11658604 on Build GRCh38
rs368576998 Chr.1: 11658625-11658625 on Build GRCh38
rs964199576 Chr.1: 11658578-11658578 on Build GRCh38
rs779809659 Chr.1: 11658585-11658585 on Build GRCh38
rs180915564 Chr.1: 11658700-11658700 on Build GRCh38
rs148817401 Chr.1: 11658762-11658762 on Build GRCh38
rs200887520 Chr.1: 11658739-11658739 on Build GRCh38
rs749796397 Chr.1: 11658694-11658694 on Build GRCh38
rs371284261 Chr.1: 11658608-11658608 on Build GRCh38
rs750684404 Chr.1: 11658644-11658644 on Build GRCh38
rs375867615 Chr.1: 11658615-11658615 on Build GRCh38
rs1003075233 Chr.1: 11658681-11658681 on Build GRCh38
rs12746243 Chr.1: 11658659-11658659 on Build GRCh38
rs754420840 Chr.1: 11658600-11658600 on Build GRCh38
rs765207374 Chr.1: 11658729-11658729 on Build GRCh38
rs142482421 Chr.1: 11658570-11658570 on Build GRCh38
rs757856396 Chr.1: 11658572-11658572 on Build GRCh38
rs752651963 Chr.1: 11658733-11658733 on Build GRCh38

View Full Product Details