Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 53823240-53823507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF2B
Entrez Gene ID
84643
Gene Name
kinesin family member 2B
Gene Aliases
-
UniGene
Hs.226805
Gene Chromosome Location
Chr.17: 53822878-53825212 on Build GRCh38
Transcripts
NM_032559.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4955427 c.299T>G p.L100R Chr.17: 53823332-53823332 on Build GRCh38
6988035 c.426G>T p.M142I Chr.17: 53823459-53823459 on Build GRCh38
3519658 c.354G>A p.M118I Chr.17: 53823387-53823387 on Build GRCh38
6856086 c.282C>A p.P94P Chr.17: 53823315-53823315 on Build GRCh38
282240 c.350C>T p.A117V Chr.17: 53823383-53823383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756381531 Chr.17: 53823480-53823480 on Build GRCh38
rs749735915 Chr.17: 53823283-53823283 on Build GRCh38
rs768138818 Chr.17: 53823444-53823444 on Build GRCh38
rs767707786 Chr.17: 53823476-53823476 on Build GRCh38
rs904138536 Chr.17: 53823459-53823459 on Build GRCh38
rs780099410 Chr.17: 53823391-53823391 on Build GRCh38
rs554238256 Chr.17: 53823266-53823266 on Build GRCh38
rs773091731 Chr.17: 53823468-53823468 on Build GRCh38
rs756888791 Chr.17: 53823269-53823269 on Build GRCh38
rs753464095 Chr.17: 53823367-53823367 on Build GRCh38
rs761543520 Chr.17: 53823467-53823467 on Build GRCh38
rs748512385 Chr.17: 53823335-53823335 on Build GRCh38
rs778212240 Chr.17: 53823423-53823423 on Build GRCh38
rs771284173 Chr.17: 53823361-53823361 on Build GRCh38
rs762624867 Chr.17: 53823470-53823470 on Build GRCh38
rs763520708 Chr.17: 53823358-53823358 on Build GRCh38
rs776054453 Chr.17: 53823462-53823462 on Build GRCh38
rs986515152 Chr.17: 53823418-53823418 on Build GRCh38
rs755087980 Chr.17: 53823402-53823402 on Build GRCh38
rs796994270 Chr.17: 53823481-53823481 on Build GRCh38
rs866206563 Chr.17: 53823379-53823379 on Build GRCh38
rs145539514 Chr.17: 53823305-53823305 on Build GRCh38
rs9912492 Chr.17: 53823415-53823415 on Build GRCh38
rs760224435 Chr.17: 53823442-53823442 on Build GRCh38
rs763682994 Chr.17: 53823273-53823273 on Build GRCh38
rs117796484 Chr.17: 53823419-53823419 on Build GRCh38
rs373634973 Chr.17: 53823433-53823433 on Build GRCh38
rs760134618 Chr.17: 53823365-53823365 on Build GRCh38
rs774804947 Chr.17: 53823438-53823438 on Build GRCh38
rs367847814 Chr.17: 53823449-53823449 on Build GRCh38
rs751532104 Chr.17: 53823474-53823474 on Build GRCh38
rs770305310 Chr.17: 53823319-53823320 on Build GRCh38
rs1043953889 Chr.17: 53823452-53823452 on Build GRCh38
rs766469120 Chr.17: 53823473-53823473 on Build GRCh38
rs769032021 Chr.17: 53823312-53823312 on Build GRCh38
rs138375059 Chr.17: 53823406-53823407 on Build GRCh38
rs758387314 Chr.17: 53823384-53823384 on Build GRCh38
rs191384582 Chr.17: 53823372-53823372 on Build GRCh38
rs747037102 Chr.17: 53823311-53823311 on Build GRCh38
rs781623963 Chr.17: 53823408-53823408 on Build GRCh38
rs59657238 Chr.17: 53823475-53823475 on Build GRCh38
rs773551999 Chr.17: 53823466-53823467 on Build GRCh38
rs115513389 Chr.17: 53823277-53823277 on Build GRCh38
rs763773013 Chr.17: 53823366-53823366 on Build GRCh38
rs372558151 Chr.17: 53823306-53823306 on Build GRCh38
rs3803824 Chr.17: 53823368-53823368 on Build GRCh38
rs774999118 Chr.17: 53823362-53823362 on Build GRCh38
rs781534412 Chr.17: 53823324-53823324 on Build GRCh38
rs750373414 Chr.17: 53823290-53823290 on Build GRCh38
rs371085430 Chr.17: 53823344-53823344 on Build GRCh38
rs771455203 Chr.17: 53823435-53823435 on Build GRCh38
rs935486867 Chr.17: 53823461-53823461 on Build GRCh38
rs17654267 Chr.17: 53823363-53823363 on Build GRCh38
rs751730942 Chr.17: 53823392-53823392 on Build GRCh38
rs144757870 Chr.17: 53823289-53823289 on Build GRCh38
rs543663752 Chr.17: 53823345-53823345 on Build GRCh38
rs757042961 Chr.17: 53823288-53823288 on Build GRCh38
rs752661021 Chr.17: 53823477-53823477 on Build GRCh38

View Full Product Details