Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43152743-43152986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM183A
Entrez Gene ID
440585
Gene Name
family with sequence similarity 183 member A
Gene Aliases
-
UniGene
Hs.657614
Gene Chromosome Location
Chr.1: 43142959-43156396 on Build GRCh38
Transcripts
NM_001101376.2 XM_005270875.4 XM_005270876.4 XM_017001322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6273012 c.281G>T p.W94L Chr.1: 43152915-43152915 on Build GRCh38
5815028 c.219T>A p.H73Q Chr.1: 43152853-43152853 on Build GRCh38
2170211 c.271G>C p.E91Q Chr.1: 43152905-43152905 on Build GRCh38
5705455 c.228G>T p.Q76H Chr.1: 43152862-43152862 on Build GRCh38
2170213 c.274G>A p.V92I Chr.1: 43152908-43152908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752690379 Chr.1: 43152884-43152884 on Build GRCh38
rs12074551 Chr.1: 43152925-43152925 on Build GRCh38
rs754797419 Chr.1: 43152870-43152870 on Build GRCh38
rs748756821 Chr.1: 43152939-43152939 on Build GRCh38
rs370598881 Chr.1: 43152835-43152835 on Build GRCh38
rs769158678 Chr.1: 43152938-43152938 on Build GRCh38
rs375131223 Chr.1: 43152879-43152879 on Build GRCh38
rs766564611 Chr.1: 43152848-43152848 on Build GRCh38
rs747989007 Chr.1: 43152802-43152802 on Build GRCh38
rs776033339 Chr.1: 43152813-43152813 on Build GRCh38
rs950217407 Chr.1: 43152766-43152766 on Build GRCh38
rs777084648 Chr.1: 43152809-43152809 on Build GRCh38
rs901905685 Chr.1: 43152954-43152954 on Build GRCh38
rs377138750 Chr.1: 43152819-43152819 on Build GRCh38
rs745450276 Chr.1: 43152937-43152937 on Build GRCh38
rs927464082 Chr.1: 43152805-43152805 on Build GRCh38
rs757083494 Chr.1: 43152892-43152893 on Build GRCh38
rs754949757 Chr.1: 43152798-43152798 on Build GRCh38
rs765154145 Chr.1: 43152904-43152904 on Build GRCh38
rs887785210 Chr.1: 43152880-43152880 on Build GRCh38
rs773390672 Chr.1: 43152961-43152961 on Build GRCh38
rs747403161 Chr.1: 43152818-43152818 on Build GRCh38
rs770698425 Chr.1: 43152812-43152812 on Build GRCh38
rs751731426 Chr.1: 43152857-43152857 on Build GRCh38
rs777405116 Chr.1: 43152901-43152901 on Build GRCh38
rs376842181 Chr.1: 43152922-43152922 on Build GRCh38
rs765416428 Chr.1: 43152830-43152830 on Build GRCh38
rs747583718 Chr.1: 43152947-43152947 on Build GRCh38
rs777202352 Chr.1: 43152823-43152823 on Build GRCh38
rs762171262 Chr.1: 43152824-43152824 on Build GRCh38
rs755989113 Chr.1: 43152888-43152888 on Build GRCh38
rs778742253 Chr.1: 43152931-43152931 on Build GRCh38
rs767230800 Chr.1: 43152873-43152873 on Build GRCh38
rs748729208 Chr.1: 43152923-43152923 on Build GRCh38
rs751334397 Chr.1: 43152779-43152779 on Build GRCh38
rs750415169 Chr.1: 43152948-43152951 on Build GRCh38
rs749040610 Chr.1: 43152810-43152810 on Build GRCh38

View Full Product Details