Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 92352227-92352458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT3
Entrez Gene ID
120114
Gene Name
FAT atypical cadherin 3
Gene Aliases
CDHF15, CDHR10, hFat3
UniGene
Hs.98523
Gene Chromosome Location
Chr.11: 92224636-92896473 on Build GRCh38
Transcripts
NM_001008781.2 XM_017017178.1 XM_017017179.1 XM_017017180.1 XM_017017181.1 XM_017017182.1 XM_017017183.1 XM_017017184.1 XM_017017186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
123066 c.122T>C p.F41S Chr.11: 92352234-92352234 on Build GRCh38
4826068 c.301G>T p.D101Y Chr.11: 92352413-92352413 on Build GRCh38
5827072 c.319A>C p.I107L Chr.11: 92352431-92352431 on Build GRCh38
4438436 c.158A>T p.Y53F Chr.11: 92352270-92352270 on Build GRCh38
5037709 c.345C>T p.A115A Chr.11: 92352457-92352457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187416225 Chr.11: 92352256-92352256 on Build GRCh38
rs571950614 Chr.11: 92352295-92352295 on Build GRCh38
rs769208873 Chr.11: 92352381-92352381 on Build GRCh38
rs200399875 Chr.11: 92352266-92352266 on Build GRCh38
rs374102428 Chr.11: 92352265-92352265 on Build GRCh38
rs372855626 Chr.11: 92352435-92352435 on Build GRCh38
rs756831292 Chr.11: 92352370-92352370 on Build GRCh38
rs749228392 Chr.11: 92352352-92352352 on Build GRCh38
rs762097164 Chr.11: 92352313-92352313 on Build GRCh38
rs778814993 Chr.11: 92352269-92352269 on Build GRCh38
rs769023260 Chr.11: 92352300-92352300 on Build GRCh38
rs1007861912 Chr.11: 92352319-92352319 on Build GRCh38
rs752550024 Chr.11: 92352345-92352345 on Build GRCh38
rs770532545 Chr.11: 92352292-92352292 on Build GRCh38
rs773530531 Chr.11: 92352412-92352412 on Build GRCh38
rs531760483 Chr.11: 92352374-92352374 on Build GRCh38
rs551585762 Chr.11: 92352421-92352421 on Build GRCh38
rs542847857 Chr.11: 92352364-92352364 on Build GRCh38
rs374727688 Chr.11: 92352296-92352296 on Build GRCh38
rs193044124 Chr.11: 92352367-92352367 on Build GRCh38
rs779334064 Chr.11: 92352385-92352385 on Build GRCh38
rs904089851 Chr.11: 92352341-92352341 on Build GRCh38
rs751323947 Chr.11: 92352368-92352368 on Build GRCh38
rs753836681 Chr.11: 92352334-92352334 on Build GRCh38
rs752495005 Chr.11: 92352270-92352270 on Build GRCh38
rs772488577 Chr.11: 92352407-92352407 on Build GRCh38
rs748640495 Chr.11: 92352397-92352397 on Build GRCh38
rs371469483 Chr.11: 92352282-92352282 on Build GRCh38
rs540982346 Chr.11: 92352331-92352331 on Build GRCh38
rs367970576 Chr.11: 92352277-92352277 on Build GRCh38
rs774809226 Chr.11: 92352309-92352309 on Build GRCh38
rs138287952 Chr.11: 92352250-92352250 on Build GRCh38
rs760878660 Chr.11: 92352324-92352324 on Build GRCh38
rs573991535 Chr.11: 92352342-92352342 on Build GRCh38
rs780805417 Chr.11: 92352373-92352373 on Build GRCh38
rs930053800 Chr.11: 92352279-92352279 on Build GRCh38
rs761003870 Chr.11: 92352422-92352422 on Build GRCh38
rs772295182 Chr.11: 92352323-92352323 on Build GRCh38
rs371896202 Chr.11: 92352264-92352264 on Build GRCh38
rs867800145 Chr.11: 92352427-92352427 on Build GRCh38
rs560453745 Chr.11: 92352337-92352337 on Build GRCh38

View Full Product Details